Search
Search
| Gene Symbol: Fstl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053629.3 | NP_446081.1 | 3 - 4 | 504 | 64 | |
| GenBank mRNA | AB021295.1 | - | 3 - 4 | 504 | 64 | |
| AB071603.1 | - | 525 | 64 | |||
Set Membership: |
|