Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586212_m1
          See other KCNH4 GE Assays ›
          Gene Symbol:
          Kcnh4
          Gene Name:
          potassium channel, voltage gated eag related subfamily H, member 4
          Gene Aliases:
          Bec2
          Chromosome Location:
          Chr.10: 88647564 - 88667382 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586212_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnh4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053630.1 NP_446082.1 15 - 16 2789 71
          GenBank mRNA AB022699.1 - 15 - 16 2789 71
          AJ007628.1 - 15 - 16 2752 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003808.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnh4

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphorelay signal transduction system
          signal transduction by protein phosphorylation
          regulation of ion transmembrane transport
          regulation of membrane potential
          potassium ion transmembrane transport
          phosphorelay sensor kinase activity
          voltage-gated potassium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P125345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238371 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121083 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11966 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170839 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125345 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P125345 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P606156 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96207 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53889 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33170 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45913 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10747466 - -
          Affymetrix Rat Gene 1.0 ST Array 10747469 - -
          Affymetrix Rat Gene 1.0 ST Array 10747481 - -
          Affymetrix Rat Gene 1.0 ST Array 10747467 - -
          Affymetrix Rat Gene 1.0 ST Array 10747480 - -
          Affymetrix Rat Gene 1.0 ST Array 10747474 - -
          Affymetrix Rat Gene 1.0 ST Array 10747478 - -
          Affymetrix Rat Gene 1.0 ST Array 10747465 - -
          Affymetrix Rat Gene 1.0 ST Array 10747471 - -
          Affymetrix Rat Gene 1.0 ST Array 10747472 - -
          Affymetrix Rat Gene 1.0 ST Array 10747476 - -
          Affymetrix Rat Gene 1.0 ST Array 10747477 - -
          Affymetrix Rat Gene 1.0 ST Array 10747473 - -
          Affymetrix Rat Gene 1.0 ST Array 10747475 - -
          Affymetrix Rat Gene 1.0 ST Array 10747479 - -
          Affymetrix Rat Gene 1.0 ST Array 10747468 - -
          Affymetrix Rat Gene 1.0 ST Array 10747470 - -
          Affymetrix Rat Gene 1.1 ST Array 10747464 - -
          Rat Genome 230 2.0 Array 1368357_PM_at - -
          Rat Genome 230 2.0 Array 1368357_at - -
          Affymetrix HT Rat Focus Array 1368357_PM_at - -
          Affymetrix HT Rat Focus Array 1368357_at - -
          Affymetrix Rat Clariom S Assay PSR1000034178.rn.1 TC1000003808.rn.1 RTA TC
          Rat Expression Set 230 1368357_PM_at - -
          Rat Expression Set 230 1368357_at - -
          Affymetrix Rat Gene 1.0 ST Array 10747464 - -
          Rat Genome U34 Set AJ007628_at - -
          Rat Genome U34 Set AJ007628_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21678642 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353444 - -
          RatRef-12 Expression BeadChip ILMN_49895 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline