Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586304_m1
          See other CDS2 GE Assays ›
          Gene Symbol:
          Cds2
          Gene Name:
          CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 124896657 - 124935221 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586304_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cds2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053643.1 NP_446095.1 10 - 11 1061 113
          GenBank mRNA AB052898.1 - 10 - 11 1061 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001767.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cds2

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          CDP-diacylglycerol biosynthetic process
          phosphatidate cytidylyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P100406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129487 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144772 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119979 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102104 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100406 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100406 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P100406 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12882 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81507 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19929 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32927 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840086 - -
          Affymetrix Rat Gene 1.0 ST Array 10840088 - -
          Affymetrix Rat Gene 1.0 ST Array 10840092 - -
          Affymetrix Rat Gene 1.0 ST Array 10840095 - -
          Affymetrix Rat Gene 1.0 ST Array 10840099 - -
          Affymetrix Rat Gene 1.0 ST Array 10840087 - -
          Affymetrix Rat Gene 1.0 ST Array 10840093 - -
          Affymetrix Rat Gene 1.0 ST Array 10840091 - -
          Affymetrix Rat Gene 1.0 ST Array 10840097 - -
          Affymetrix Rat Gene 1.0 ST Array 10840089 - -
          Affymetrix Rat Gene 1.0 ST Array 10840096 - -
          Affymetrix Rat Gene 1.0 ST Array 10840098 - -
          Affymetrix Rat Gene 1.0 ST Array 10840090 - -
          Affymetrix Rat Gene 1.0 ST Array 10840094 - -
          Affymetrix Rat Gene 1.1 ST Array 10840085 - -
          Rat Genome 230 2.0 Array 1369505_PM_at - -
          Rat Genome 230 2.0 Array 1369505_at - -
          Affymetrix HT Rat Focus Array 1369505_PM_at - -
          Affymetrix HT Rat Focus Array 1369505_at - -
          Affymetrix Rat Clariom S Assay PSR0300012781.rn.1 TC0300001767.rn.1 RTA TC
          Rat Expression Set 230 1369505_at - -
          Rat Expression Set 230 1369505_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840085 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20861610 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357772 - -
          RatRef-12 Expression BeadChip ILMN_68503 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline