Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586491_m1
          See other P2RX2 GE Assays ›
          Gene Symbol:
          P2rx2
          Gene Name:
          purinergic receptor P2X, ligand-gated ion channel, 2
          Gene Aliases:
          P2X2
          Chromosome Location:
          Chr.12: 52397654 - 52401005 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586491_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2rx2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053656.2 NP_446108.2 7 - 8 776 64
          XM_006249515.2 XP_006249577.1 7 - 8 969 64
          XM_006249516.2 XP_006249578.1 7 - 8 968 64
          XM_006249517.2 XP_006249579.1 7 - 8 931 64
          XM_008769305.1 XP_008767527.1 6 - 7 852 64
          XM_008769306.1 XP_008767528.1 7 - 8 645 64
          XM_008769307.1 XP_008767529.1 5 - 6 717 64
          GenBank mRNA AF020756.1 - 6 - 7 676 64
          AF020757.1 - 6 - 7 722 64
          AF020758.1 - 5 - 6 816 64
          AF020759.1 - 7 - 8 738 64
          AF064549.1 - 6 - 7 813 64
          U14414.1 - 7 - 8 776 64
          Y10473.1 - 7 - 8 776 64
          Y10474.1 - 7 - 8 758 64
          Y10475.1 - 7 - 8 812 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000878.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2rx2

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          detection of hypoxic conditions in blood by carotid body chemoreceptor signaling
          cation transport
          chemical synaptic transmission
          neuromuscular synaptic transmission
          neuromuscular junction development
          sensory perception of sound
          response to carbohydrate
          response to organic substance
          urinary bladder smooth muscle contraction
          neuronal action potential
          peristalsis
          response to ATP
          ion transmembrane transport
          purinergic nucleotide receptor signaling pathway
          behavioral response to pain
          skeletal muscle fiber development
          sensory perception of taste
          protein homooligomerization
          protein heterooligomerization
          cation transmembrane transport
          purinergic nucleotide receptor activity
          extracellular ATP-gated cation channel activity
          ion channel activity
          copper ion binding
          protein binding
          ATP binding
          drug binding
          zinc ion binding
          ligand-gated ion channel activity
          nickel cation binding
          phosphatidylinositol binding
          identical protein binding
          mercury ion binding
          cadmium ion binding
          cobalt ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P288091 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3628 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138509 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288091 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138509 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288091 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138509 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P288091 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288091 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P512017 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10759401 - -
          Affymetrix Rat Gene 1.0 ST Array 10759395 - -
          Affymetrix Rat Gene 1.0 ST Array 10759398 - -
          Affymetrix Rat Gene 1.0 ST Array 10759388 - -
          Affymetrix Rat Gene 1.0 ST Array 10759387 - -
          Affymetrix Rat Gene 1.0 ST Array 10759394 - -
          Affymetrix Rat Gene 1.0 ST Array 10759396 - -
          Affymetrix Rat Gene 1.0 ST Array 10759384 - -
          Affymetrix Rat Gene 1.0 ST Array 10759397 - -
          Affymetrix Rat Gene 1.0 ST Array 10759399 - -
          Affymetrix Rat Gene 1.0 ST Array 10759393 - -
          Affymetrix Rat Gene 1.0 ST Array 10759386 - -
          Affymetrix Rat Gene 1.0 ST Array 10759391 - -
          Affymetrix Rat Gene 1.0 ST Array 10759389 - -
          Affymetrix Rat Gene 1.0 ST Array 10759400 - -
          Affymetrix Rat Gene 1.0 ST Array 10759392 - -
          Affymetrix Rat Gene 1.0 ST Array 10759390 - -
          Affymetrix Rat Gene 1.1 ST Array 10759383 - -
          Rat Genome 230 2.0 Array 1369474_a_at - -
          Rat Genome 230 2.0 Array 1369474_PM_a_at - -
          Rat Genome 230 2.0 Array 1387578_PM_a_at - -
          Rat Genome 230 2.0 Array 1369475_PM_x_at - -
          Rat Genome 230 2.0 Array 1369475_x_at - -
          Rat Genome 230 2.0 Array 1387578_a_at - -
          Affymetrix HT Rat Focus Array 1387578_a_at - -
          Affymetrix HT Rat Focus Array 1369474_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387578_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369474_a_at - -
          Affymetrix HT Rat Focus Array 1369475_PM_x_at - -
          Affymetrix HT Rat Focus Array 1369475_x_at - -
          Affymetrix Rat Clariom S Assay PSR1200007789.rn.1 TC1200000878.rn.1 RTA TC
          Rat Expression Set 230 1369475_x_at - -
          Rat Expression Set 230 1369474_a_at - -
          Rat Expression Set 230 1369474_PM_a_at - -
          Rat Expression Set 230 1387578_a_at - -
          Rat Expression Set 230 1369475_PM_x_at - -
          Rat Expression Set 230 1387578_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10759383 - -
          Rat Genome U34 Set AF020758_g_at - -
          Rat Genome U34 Set AF020757_s_at - -
          Rat Genome U34 Set U14414_at - -
          Rat Genome U34 Set AF020756_s_at - -
          Rat Genome U34 Set AF028603_s_at - -
          Rat Genome U34 Set AF020758_at - -
          Rat Genome U34 Set AF028603_s_at - -
          Rat Genome U34 Set AF020758_g_at - -
          Rat Genome U34 Set AF020757_s_at - -
          Rat Genome U34 Set AF020756_s_at - -
          Rat Genome U34 Set U14414_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21282547 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354323 - -
          RatRef-12 Expression BeadChip ILMN_58643 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline