Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586578_m1
          See other P3H1 GE Assays ›
          Gene Symbol:
          P3h1
          Gene Name:
          prolyl 3-hydroxylase 1
          Gene Aliases:
          Gros1, Lepre1
          Chromosome Location:
          Chr.5: 138279597 - 138294280 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586578_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P3h1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053667.1 NP_446119.1 5 - 6 1076 58
          GenBank mRNA AF087433.1 - 5 - 6 1076 58
          BC161838.1 - 5 - 6 1110 9101602 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001347.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P3h1

          Panther Classification:

          Molecular Function -

          extracellular matrix glycoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          cell growth
          protein hydroxylation
          peptidyl-proline hydroxylation
          collagen fibril organization
          regulation of ossification
          regulation of protein secretion
          protein stabilization
          oxidation-reduction process
          bone development
          negative regulation of post-translational protein modification
          iron ion binding
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
          procollagen-proline 3-dioxygenase activity
          L-ascorbic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P059217 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P097814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88668 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142104 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059217 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097814 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097814 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059217 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P097814 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P059217 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P498149 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701318161 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25223 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5103 - -
          Agilent Rat cDNA Microarray (G4105A) 701318161 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10871544 - -
          Affymetrix Rat Gene 1.0 ST Array 10871545 - -
          Affymetrix Rat Gene 1.0 ST Array 10871547 - -
          Affymetrix Rat Gene 1.0 ST Array 10871557 - -
          Affymetrix Rat Gene 1.0 ST Array 10871553 - -
          Affymetrix Rat Gene 1.0 ST Array 10871552 - -
          Affymetrix Rat Gene 1.0 ST Array 10871542 - -
          Affymetrix Rat Gene 1.0 ST Array 10871559 - -
          Affymetrix Rat Gene 1.0 ST Array 10871549 - -
          Affymetrix Rat Gene 1.0 ST Array 10871541 - -
          Affymetrix Rat Gene 1.0 ST Array 10871560 - -
          Affymetrix Rat Gene 1.0 ST Array 10871554 - -
          Affymetrix Rat Gene 1.0 ST Array 10871556 - -
          Affymetrix Rat Gene 1.0 ST Array 10871548 - -
          Affymetrix Rat Gene 1.0 ST Array 10871546 - -
          Affymetrix Rat Gene 1.0 ST Array 10871551 - -
          Affymetrix Rat Gene 1.0 ST Array 10871543 - -
          Affymetrix Rat Gene 1.0 ST Array 10871550 - -
          Affymetrix Rat Gene 1.0 ST Array 10871558 - -
          Affymetrix Rat Gene 1.1 ST Array 10871540 - -
          Rat Genome 230 2.0 Array 1367967_at - -
          Rat Genome 230 2.0 Array 1367967_PM_at - -
          Affymetrix HT Rat Focus Array 1367967_at - -
          Affymetrix HT Rat Focus Array 1367967_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500009384.rn.1 TC0500001347.rn.1 RTA TC
          Rat Expression Set 230 1367967_at - -
          Rat Expression Set 230 1367967_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10871540 - -
          Rat Genome U34 Set rc_AI007812_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21414692 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361231 - -
          RatRef-12 Expression BeadChip ILMN_58084 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline