Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586595_m1
          See other TMF1 GE Assays ›
          Gene Symbol:
          Tmf1
          Gene Name:
          TATA element modulatory factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 129521986 - 129546028 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586595_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053671.2 NP_446123.2 2 - 3 1421 73
          XM_008763143.1 XP_008761365.1 2 - 3 2943 73
          GenBank mRNA AF107843.1 - 2 - 3 1344 73
          CK473511.1 - 1 - 2 273 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400004163.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmf1

          Gene Ontology Categories:

          Function(s)

          acrosome assembly
          positive regulation of cytokine production
          spermatid development
          spermatid nucleus differentiation
          male gonad development
          negative regulation of gene expression
          sperm motility
          luteinizing hormone secretion
          Leydig cell differentiation
          defense response to bacterium
          negative regulation of apoptotic process
          regulation of proteasomal protein catabolic process
          cellular response to organic cyclic compound
          positive regulation of testosterone secretion

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P444247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185659 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96408 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100347 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P444247 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P444247 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P444247 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P444247 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701224165 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44573 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28807 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80979 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11309 - -
          Agilent Rat cDNA Microarray (G4105A) 701224165 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10864354 - -
          Affymetrix Rat Gene 1.0 ST Array 10864355 - -
          Affymetrix Rat Gene 1.0 ST Array 10864353 - -
          Affymetrix Rat Gene 1.0 ST Array 10864352 - -
          Affymetrix Rat Gene 1.1 ST Array 10864351 - -
          Rat Genome 230 2.0 Array 1387350_PM_at - -
          Rat Genome 230 2.0 Array 1399066_at - -
          Rat Genome 230 2.0 Array 1399066_PM_at - -
          Rat Genome 230 2.0 Array 1387350_at - -
          Affymetrix HT Rat Focus Array 1399066_at - -
          Affymetrix HT Rat Focus Array 1387350_PM_at - -
          Affymetrix HT Rat Focus Array 1387350_at - -
          Affymetrix HT Rat Focus Array 1399066_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400029949.rn.1 TC0400004163.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029951.rn.1 TC0400004163.rn.1 RTA TC
          Rat Expression Set 230 1387350_at - -
          Rat Expression Set 230 1387350_PM_at - -
          Rat Expression Set 230 1399066_at - -
          Rat Expression Set 230 1399066_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10864351 - -
          Rat Genome U34 Set rc_AI045428_at - -
          Rat Genome U34 Set rc_AI236763_at - -
          Rat Genome U34 Set rc_AI175610_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21783146 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline