Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586609_m1
          See other PHYH GE Assays ›
          Gene Symbol:
          Phyh
          Gene Name:
          phytanoyl-CoA 2-hydroxylase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 77287580 - 77304482 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586609_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phyh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053674.1 NP_446126.1 7 - 8 839 62
          GenBank mRNA AF121345.1 - 7 - 8 839 62
          BC086573.1 - 7 - 8 834 7306754 62
          FQ216943.1 - 5 - 6 671 62
          FQ224924.1 - 5 - 6 671 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700002211.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Phyh

          Panther Classification:

          Molecular Function -

          hydroxylase

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid alpha-oxidation
          isoprenoid metabolic process
          methyl-branched fatty acid metabolic process
          catalytic activity
          L-ascorbic acid binding
          metal ion binding
          cofactor binding
          phytanoyl-CoA dioxygenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P191021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118513 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28560 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P191021 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P191021 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P191021 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P191021 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P191022 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700641917 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84386 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102594 - -
          Agilent Rat cDNA Microarray (G4105A) 700641917 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10799563 - -
          Affymetrix Rat Gene 1.0 ST Array 10799567 - -
          Affymetrix Rat Gene 1.0 ST Array 10799565 - -
          Affymetrix Rat Gene 1.0 ST Array 10799566 - -
          Affymetrix Rat Gene 1.0 ST Array 10799564 - -
          Affymetrix Rat Gene 1.0 ST Array 10799559 - -
          Affymetrix Rat Gene 1.0 ST Array 10799561 - -
          Affymetrix Rat Gene 1.0 ST Array 10799560 - -
          Affymetrix Rat Gene 1.0 ST Array 10799562 - -
          Affymetrix Rat Gene 1.1 ST Array 10799558 - -
          Rat Genome 230 2.0 Array 1393245_at - -
          Rat Genome 230 2.0 Array 1387271_at - -
          Rat Genome 230 2.0 Array 1387271_PM_at - -
          Rat Genome 230 2.0 Array 1393245_PM_at - -
          Affymetrix HT Rat Focus Array 1387271_PM_at - -
          Affymetrix HT Rat Focus Array 1393245_at - -
          Affymetrix HT Rat Focus Array 1387271_at - -
          Affymetrix HT Rat Focus Array 1393245_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700014756.rn.1 TC1700002211.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700014754.rn.1 TC1700002211.rn.1 RTA TC
          Rat Expression Set 230 1387271_PM_at - -
          Rat Expression Set 230 1387271_at - -
          Rat Expression Set 230 1393245_PM_at - -
          Rat Expression Set 230 1393245_at - -
          Affymetrix Rat Gene 1.0 ST Array 10799558 - -
          Rat Genome U34 Set rc_AA899815_at - -
          Rat Genome U34 Set rc_AI029057_at - -
          Rat Genome U34 Set rc_AI172143_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22329996 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350834 - -
          RatRef-12 Expression BeadChip ILMN_56893 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline