Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586652_m1
          See other NOX1 GE Assays ›
          Gene Symbol:
          Nox1
          Gene Name:
          NADPH oxidase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 104909328 - 104932508 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586652_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nox1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053683.1 NP_446135.1 12 - 13 1691 82
          GenBank mRNA AF152963.1 - 12 - 13 1691 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002179.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nox1

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          angiogenesis
          regulation of systemic arterial blood pressure by renin-angiotensin
          superoxide metabolic process
          signal transduction
          positive regulation of cell proliferation
          positive regulation of vascular endothelial growth factor production
          proton transport
          cell migration
          extracellular matrix organization
          superoxide anion generation
          hydrogen peroxide metabolic process
          positive regulation of MAPK cascade
          positive regulation of integrin biosynthetic process
          positive regulation of JNK cascade
          positive regulation of smooth muscle cell proliferation
          intracellular pH elevation
          oxidation-reduction process
          cellular response to hyperoxia
          oxygen metabolic process
          positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          cellular stress response to acidic pH
          positive regulation of reactive oxygen species metabolic process
          protein binding
          superoxide-generating NADPH oxidase activity
          voltage-gated proton channel activity
          metal ion binding
          Rac GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P091732 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091732 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091732 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P091732 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P421322 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940550 - -
          Affymetrix Rat Gene 1.1 ST Array 10940550 - -
          Rat Genome 230 2.0 Array 1387311_PM_at - -
          Rat Genome 230 2.0 Array 1387311_at - -
          Affymetrix HT Rat Focus Array 1387311_PM_at - -
          Affymetrix HT Rat Focus Array 1387311_at - -
          Affymetrix Rat Clariom S Assay PSR0X00013603.rn.1 TC0X00002179.rn.1 RTA TC
          Rat Expression Set 230 1387311_at - -
          Rat Expression Set 230 1387311_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940550 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21896757 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline