Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586715_m1
          See other CCL28 GE Assays ›
          Gene Symbol:
          Ccl28
          Gene Name:
          chemokine (C-C motif) ligand 28
          Gene Aliases:
          Scya28
          Chromosome Location:
          Chr.2: 52379341 - 52403979 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586715_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccl28
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053700.1 NP_446152.1 1 - 2 107 66
          GenBank mRNA AF361490.1 - 1 - 2 107 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000468.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Rat Chemokine Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccl28

          Panther Classification:

          Molecular Function -

          chemokine

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell-matrix adhesion
          immune response
          positive regulation of cytosolic calcium ion concentration
          response to nutrient
          cell chemotaxis
          negative regulation of leukocyte tethering or rolling
          molecular_function
          chemokine activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138695 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143501 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157778 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157778 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157778 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12903 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64392 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15280 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813211 - -
          Affymetrix Rat Gene 1.0 ST Array 10813212 - -
          Affymetrix Rat Gene 1.0 ST Array 10813213 - -
          Affymetrix Rat Gene 1.1 ST Array 10813210 - -
          Rat Genome 230 2.0 Array 1369513_at - -
          Rat Genome 230 2.0 Array 1369513_PM_at - -
          Affymetrix HT Rat Focus Array 1369513_PM_at - -
          Affymetrix HT Rat Focus Array 1369513_at - -
          Affymetrix Rat Clariom S Assay PSR0200002691.rn.1 TC0200000468.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200002690.rn.1 TC0200000468.rn.1 RTA TC
          Rat Expression Set 230 1369513_PM_at - -
          Rat Expression Set 230 1369513_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813210 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21527447 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364159 - -
          RatRef-12 Expression BeadChip ILMN_51980 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline