Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586734_m1
          See other CACNA1F GE Assays ›
          Gene Symbol:
          Cacna1f
          Gene Name:
          calcium channel, voltage-dependent, L type, alpha 1F subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 15712709 - 15741099 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586734_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cacna1f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053701.1 NP_446153.1 27 - 28 3294 65
          XM_006256698.2 XP_006256760.1 23 - 24 2671 65
          GenBank mRNA AF365975.1 - 27 - 28 3294 65
          DQ393415.1 - 27 - 28 3298 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001508.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cacna1f

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular calcium ion homeostasis
          neurotransmitter secretion
          axonogenesis
          visual perception
          regulation of ion transmembrane transport
          T cell homeostasis
          dendrite morphogenesis
          regulation of T cell receptor signaling pathway
          detection of light stimulus involved in visual perception
          retina development in camera-type eye
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during action potential
          voltage-gated calcium channel activity
          high voltage-gated calcium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866706 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76989 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26365 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12904 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12904 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12904 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12904 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23465 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14089 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937116 - -
          Affymetrix Rat Gene 1.0 ST Array 10937123 - -
          Affymetrix Rat Gene 1.0 ST Array 10937150 - -
          Affymetrix Rat Gene 1.0 ST Array 10937122 - -
          Affymetrix Rat Gene 1.0 ST Array 10937149 - -
          Affymetrix Rat Gene 1.0 ST Array 10937118 - -
          Affymetrix Rat Gene 1.0 ST Array 10937119 - -
          Affymetrix Rat Gene 1.0 ST Array 10937130 - -
          Affymetrix Rat Gene 1.0 ST Array 10937136 - -
          Affymetrix Rat Gene 1.0 ST Array 10937157 - -
          Affymetrix Rat Gene 1.0 ST Array 10937145 - -
          Affymetrix Rat Gene 1.0 ST Array 10937148 - -
          Affymetrix Rat Gene 1.0 ST Array 10937146 - -
          Affymetrix Rat Gene 1.0 ST Array 10937111 - -
          Affymetrix Rat Gene 1.0 ST Array 10937121 - -
          Affymetrix Rat Gene 1.0 ST Array 10937159 - -
          Affymetrix Rat Gene 1.0 ST Array 10937153 - -
          Affymetrix Rat Gene 1.0 ST Array 10937133 - -
          Affymetrix Rat Gene 1.0 ST Array 10937140 - -
          Affymetrix Rat Gene 1.0 ST Array 10937112 - -
          Affymetrix Rat Gene 1.0 ST Array 10937151 - -
          Affymetrix Rat Gene 1.0 ST Array 10937114 - -
          Affymetrix Rat Gene 1.0 ST Array 10937154 - -
          Affymetrix Rat Gene 1.0 ST Array 10937126 - -
          Affymetrix Rat Gene 1.0 ST Array 10937131 - -
          Affymetrix Rat Gene 1.0 ST Array 10937138 - -
          Affymetrix Rat Gene 1.0 ST Array 10937134 - -
          Affymetrix Rat Gene 1.0 ST Array 10937125 - -
          Affymetrix Rat Gene 1.0 ST Array 10937147 - -
          Affymetrix Rat Gene 1.0 ST Array 10937155 - -
          Affymetrix Rat Gene 1.0 ST Array 10937120 - -
          Affymetrix Rat Gene 1.0 ST Array 10937152 - -
          Affymetrix Rat Gene 1.0 ST Array 10937137 - -
          Affymetrix Rat Gene 1.0 ST Array 10937127 - -
          Affymetrix Rat Gene 1.0 ST Array 10937113 - -
          Affymetrix Rat Gene 1.0 ST Array 10937144 - -
          Affymetrix Rat Gene 1.0 ST Array 10937142 - -
          Affymetrix Rat Gene 1.0 ST Array 10937115 - -
          Affymetrix Rat Gene 1.0 ST Array 10937117 - -
          Affymetrix Rat Gene 1.0 ST Array 10937132 - -
          Affymetrix Rat Gene 1.0 ST Array 10937156 - -
          Affymetrix Rat Gene 1.0 ST Array 10937143 - -
          Affymetrix Rat Gene 1.0 ST Array 10937128 - -
          Affymetrix Rat Gene 1.0 ST Array 10937141 - -
          Affymetrix Rat Gene 1.0 ST Array 10937139 - -
          Affymetrix Rat Gene 1.0 ST Array 10937135 - -
          Affymetrix Rat Gene 1.0 ST Array 10937124 - -
          Affymetrix Rat Gene 1.0 ST Array 10937129 - -
          Affymetrix Rat Gene 1.1 ST Array 10937110 - -
          Rat Genome 230 2.0 Array 1387846_PM_at - -
          Rat Genome 230 2.0 Array 1387846_at - -
          Affymetrix HT Rat Focus Array 1387846_at - -
          Affymetrix HT Rat Focus Array 1387846_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00009710.rn.1 TC0X00001508.rn.1 RTA TC
          Rat Expression Set 230 1387846_PM_at - -
          Rat Expression Set 230 1387846_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937110 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20839387 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline