Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586815_m1
          See other EMB GE Assays ›
          Gene Symbol:
          Emb
          Gene Name:
          embigin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 49682763 - 49733475 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586815_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Emb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053719.1 NP_446171.1 5 - 6 688 144
          GenBank mRNA AJ009698.1 - 5 - 6 688 144
          BC061846.1 - 5 - 6 690 5621074 144
          FQ213962.1 - 5 - 6 701 144
          FQ228768.1 - 5 - 6 701 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000448.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Emb

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          plasma membrane lactate transport
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P304220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155862 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171304 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117279 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175061 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90434 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P304220 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P304220 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P304220 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P304220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78622 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35550 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49172 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813157 - -
          Affymetrix Rat Gene 1.0 ST Array 10813159 - -
          Affymetrix Rat Gene 1.0 ST Array 10813160 - -
          Affymetrix Rat Gene 1.0 ST Array 10813158 - -
          Affymetrix Rat Gene 1.0 ST Array 10813155 - -
          Affymetrix Rat Gene 1.0 ST Array 10813153 - -
          Affymetrix Rat Gene 1.0 ST Array 10813154 - -
          Affymetrix Rat Gene 1.0 ST Array 10813161 - -
          Affymetrix Rat Gene 1.0 ST Array 10813156 - -
          Affymetrix Rat Gene 1.0 ST Array 10821541 - -
          Affymetrix Rat Gene 1.1 ST Array 10813152 - -
          Rat Genome 230 2.0 Array 1368541_at - -
          Rat Genome 230 2.0 Array 1368541_PM_at - -
          Affymetrix HT Rat Focus Array 1368541_at - -
          Affymetrix HT Rat Focus Array 1368541_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200002599.rn.1 TC0200000448.rn.1 RTA TC
          Rat Expression Set 230 1368541_PM_at - -
          Rat Expression Set 230 1368541_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813152 - -
          Rat Genome U34 Set AJ009698_at - -
          Rat Genome U34 Set AJ009698_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21676545 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359464 - -
          RatRef-12 Expression BeadChip ILMN_58897 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline