Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586837_m1
          See other RASGRF2 GE Assays ›
          Gene Symbol:
          Rasgrf2
          Gene Name:
          RAS protein-specific guanine nucleotide-releasing factor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 21460041 - 21698937 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586837_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rasgrf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053721.1 NP_446173.1 17 - 18 2548 74
          GenBank mRNA AJ276774.1 - 17 - 18 2548 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002599.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rasgrf2

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          biological_process
          long-term synaptic potentiation
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          molecular_function
          Rho guanyl-nucleotide exchange factor activity
          calmodulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P121308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31992 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61521 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44253 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93856 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25810 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44601 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178176 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133137 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52900 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73665 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161807 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19296 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20467 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49925 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26987 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P121308 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P121308 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P121308 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P121308 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10390 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69383 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13197 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40113 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20709 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31722 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15383 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19537 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39621 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5310 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31668 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77962 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37541 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42969 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6820 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68692 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10820344 - -
          Affymetrix Rat Gene 1.0 ST Array 10820348 - -
          Affymetrix Rat Gene 1.0 ST Array 10820347 - -
          Affymetrix Rat Gene 1.0 ST Array 10820357 - -
          Affymetrix Rat Gene 1.0 ST Array 10820343 - -
          Affymetrix Rat Gene 1.0 ST Array 10820350 - -
          Affymetrix Rat Gene 1.0 ST Array 10820356 - -
          Affymetrix Rat Gene 1.0 ST Array 10820355 - -
          Affymetrix Rat Gene 1.0 ST Array 10820337 - -
          Affymetrix Rat Gene 1.0 ST Array 10820342 - -
          Affymetrix Rat Gene 1.0 ST Array 10820341 - -
          Affymetrix Rat Gene 1.0 ST Array 10820339 - -
          Affymetrix Rat Gene 1.0 ST Array 10820351 - -
          Affymetrix Rat Gene 1.0 ST Array 10820358 - -
          Affymetrix Rat Gene 1.0 ST Array 10820340 - -
          Affymetrix Rat Gene 1.0 ST Array 10820359 - -
          Affymetrix Rat Gene 1.0 ST Array 10820354 - -
          Affymetrix Rat Gene 1.0 ST Array 10820361 - -
          Affymetrix Rat Gene 1.0 ST Array 10820360 - -
          Affymetrix Rat Gene 1.0 ST Array 10820349 - -
          Affymetrix Rat Gene 1.0 ST Array 10820352 - -
          Affymetrix Rat Gene 1.0 ST Array 10820338 - -
          Affymetrix Rat Gene 1.1 ST Array 10820335 - -
          Rat Genome 230 2.0 Array 1391226_PM_at - -
          Rat Genome 230 2.0 Array 1391226_at - -
          Rat Genome 230 2.0 Array 1369780_PM_at - -
          Rat Genome 230 2.0 Array 1369780_at - -
          Affymetrix HT Rat Focus Array 1369780_at - -
          Affymetrix HT Rat Focus Array 1391226_PM_at - -
          Affymetrix HT Rat Focus Array 1391226_at - -
          Affymetrix HT Rat Focus Array 1369780_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200018086.rn.1 TC0200002599.rn.1 RTA TC
          Rat Expression Set 230 1369780_at - -
          Rat Expression Set 230 1369780_PM_at - -
          Rat Expression Set 230 1391226_at - -
          Rat Expression Set 230 1391226_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10820335 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21337413 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364232 - -
          RatRef-12 Expression BeadChip ILMN_62709 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline