Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586945_m1
          See other NCF1 GE Assays ›
          Gene Symbol:
          Ncf1
          Gene Name:
          neutrophil cytosolic factor 1
          Gene Aliases:
          Ncf-1, p47phox
          Chromosome Location:
          Chr.12: 25497104 - 25506300 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586945_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ncf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053734.2 NP_446186.2 7 - 8 697 105
          GenBank mRNA AF260779.1 - 6 - 7 625 105
          AF547392.1 - 7 - 8 697 105
          AF547393.1 - 7 - 8 697 105
          AY029167.1 - 7 - 8 697 105
          BC061810.1 - 7 - 8 697 5598653 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000452.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ncf1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to yeast
          leukocyte mediated cytotoxicity
          respiratory burst involved in defense response
          protein targeting to membrane
          leukotriene metabolic process
          apoptotic process
          inflammatory response
          cellular defense response
          cell proliferation
          response to bacterium
          superoxide anion generation
          defense response to bacterium
          respiratory burst
          negative regulation of smooth muscle contraction
          hydrogen peroxide biosynthetic process
          defense response to Gram-positive bacterium
          defense response to fungus
          oxidation-reduction process
          neutrophil mediated killing of gram-positive bacterium
          neutrophil mediated killing of fungus
          superoxide-generating NADPH oxidase activity
          SH3 domain binding
          phosphatidylinositol binding
          phosphatidylinositol-3,4-bisphosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P051322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638987 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57790 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143203 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051322 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051322 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P051322 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P298049 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58604 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10707 - -
          Agilent Rat cDNA Microarray (G4105A) 701469622 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757900 - -
          Affymetrix Rat Gene 1.0 ST Array 10757908 - -
          Affymetrix Rat Gene 1.0 ST Array 10757906 - -
          Affymetrix Rat Gene 1.0 ST Array 10757911 - -
          Affymetrix Rat Gene 1.0 ST Array 10757899 - -
          Affymetrix Rat Gene 1.0 ST Array 10757901 - -
          Affymetrix Rat Gene 1.0 ST Array 10757905 - -
          Affymetrix Rat Gene 1.0 ST Array 10757904 - -
          Affymetrix Rat Gene 1.0 ST Array 10757907 - -
          Affymetrix Rat Gene 1.0 ST Array 10757902 - -
          Affymetrix Rat Gene 1.0 ST Array 10757910 - -
          Affymetrix Rat Gene 1.0 ST Array 10757909 - -
          Affymetrix Rat Gene 1.1 ST Array 10757898 - -
          Rat Genome 230 2.0 Array 1387413_at - -
          Rat Genome 230 2.0 Array 1387413_PM_at - -
          Affymetrix HT Rat Focus Array 1387413_at - -
          Affymetrix HT Rat Focus Array 1387413_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200004137.rn.1 TC1200000452.rn.1 RTA TC
          Rat Expression Set 230 1387413_at - -
          Rat Expression Set 230 1387413_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757898 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21546613 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355786 - -
          RatRef-12 Expression BeadChip ILMN_62032 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline