Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586951_m1
          See other PI4K2A GE Assays ›
          Gene Symbol:
          Pi4k2a
          Gene Name:
          phosphatidylinositol 4-kinase type 2 alpha
          Gene Aliases:
          Pi4KII
          Chromosome Location:
          Chr.1: 261337594 - 261362509 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586951_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pi4k2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053735.1 NP_446187.1 4 - 5 977 98
          GenBank mRNA AY029199.1 - 4 - 5 977 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003632.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pi4k2a

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          basophil degranulation
          phosphatidylinositol biosynthetic process
          phosphatidylinositol phosphorylation
          1-phosphatidylinositol 4-kinase activity
          ATP binding
          protein complex binding
          AP-3 adaptor complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P128887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960242 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167654 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59996 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128887 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128887 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P128887 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P833613 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28651 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34515 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83734 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84449 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730321 - -
          Affymetrix Rat Gene 1.0 ST Array 10730316 - -
          Affymetrix Rat Gene 1.0 ST Array 10730318 - -
          Affymetrix Rat Gene 1.0 ST Array 10730319 - -
          Affymetrix Rat Gene 1.0 ST Array 10730313 - -
          Affymetrix Rat Gene 1.0 ST Array 10730317 - -
          Affymetrix Rat Gene 1.0 ST Array 10730320 - -
          Affymetrix Rat Gene 1.0 ST Array 10730314 - -
          Affymetrix Rat Gene 1.0 ST Array 10730315 - -
          Affymetrix Rat Gene 1.1 ST Array 10730312 - -
          Rat Genome 230 2.0 Array 1369177_PM_at - -
          Rat Genome 230 2.0 Array 1375272_PM_at - -
          Rat Genome 230 2.0 Array 1375272_at - -
          Rat Genome 230 2.0 Array 1369177_at - -
          Affymetrix HT Rat Focus Array 1369177_PM_at - -
          Affymetrix HT Rat Focus Array 1369177_at - -
          Affymetrix Rat Clariom S Assay PSR0100029575.rn.1 TC0100003632.rn.1 RTA TC
          Rat Expression Set 230 1369177_PM_at - -
          Rat Expression Set 230 1369177_at - -
          Affymetrix Rat Gene 1.0 ST Array 10730312 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21339106 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353104 - -
          RatRef-12 Expression BeadChip ILMN_54415 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline