Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587010_m1
          See other CDC37 GE Assays ›
          Gene Symbol:
          Cdc37
          Gene Name:
          cell division cycle 37
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 22160922 - 22173797 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587010_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdc37
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053743.1 NP_446195.1 7 - 8 1070 74
          GenBank mRNA AB097113.1 - 7 - 8 983 74
          BC061720.1 - 7 - 8 1039 5598479 74
          D26564.1 - 7 - 8 1070 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002064.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdc37

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          protein stabilization
          regulation of cell cycle
          regulation of interferon-gamma-mediated signaling pathway
          regulation of type I interferon-mediated signaling pathway
          protein C-terminus binding
          kinase binding
          protein kinase binding
          heat shock protein binding
          mitogen-activated protein kinase kinase kinase binding
          protein kinase B binding
          unfolded protein binding
          chaperone binding
          Hsp90 protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152242 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P147308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116807 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8208 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158650 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147308 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152242 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152242 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147308 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152242 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P147308 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P441773 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700325601 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58619 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37900 - -
          Agilent Rat cDNA Microarray (G4105A) 700325601 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10915546 - -
          Affymetrix Rat Gene 1.0 ST Array 10915543 - -
          Affymetrix Rat Gene 1.0 ST Array 10915542 - -
          Affymetrix Rat Gene 1.0 ST Array 10915548 - -
          Affymetrix Rat Gene 1.0 ST Array 10915544 - -
          Affymetrix Rat Gene 1.0 ST Array 10915545 - -
          Affymetrix Rat Gene 1.0 ST Array 10915541 - -
          Affymetrix Rat Gene 1.0 ST Array 10915547 - -
          Affymetrix Rat Gene 1.0 ST Array 10915540 - -
          Affymetrix Rat Gene 1.0 ST Array 10915539 - -
          Affymetrix Rat Gene 1.1 ST Array 10915538 - -
          Rat Genome 230 2.0 Array 1367453_PM_at - -
          Rat Genome 230 2.0 Array 1367453_at - -
          Affymetrix HT Rat Focus Array 1367453_PM_at - -
          Affymetrix HT Rat Focus Array 1367453_at - -
          Affymetrix Rat Clariom S Assay PSR0800016608.rn.1 TC0800002064.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800016607.rn.1 TC0800002064.rn.1 RTA TC
          Rat Expression Set 230 1367453_at - -
          Rat Expression Set 230 1367453_PM_at - -
          Rat Expression Set 230 1367453_at - -
          Rat Expression Set 230 1367453_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10915538 - -
          Rat Genome U34 Set D26564_g_at - -
          Rat Genome U34 Set D26564_at - -
          Rat Genome U34 Set rc_AI234952_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21383221 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375218 - -
          RatRef-12 Expression BeadChip ILMN_67822 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline