Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587389_m1
          See other F11R GE Assays ›
          Gene Symbol:
          F11r
          Gene Name:
          F11 receptor
          Gene Aliases:
          Jam1
          Chromosome Location:
          Chr.13: 89826272 - 89850151 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587389_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: F11r
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053796.1 NP_446248.1 1 - 2 129 98
          GenBank mRNA AF276998.1 - 1 - 2 129 98
          BC065309.1 - 1 - 2 111 5599898 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000765.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene F11r

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          response to radiation
          viral process
          cell differentiation
          epithelial cell differentiation
          actomyosin structure organization
          positive regulation of blood pressure
          intestinal absorption
          establishment of protein localization to plasma membrane
          establishment of endothelial intestinal barrier
          regulation of membrane permeability
          regulation of actin cytoskeleton reorganization
          protein binding
          PDZ domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P482595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325650 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166615 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132580 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165846 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P482595 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P482595 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P482595 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P482595 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700482568 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7868 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27649 - -
          Agilent Rat cDNA Microarray (G4105A) 700482568 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10765601 - -
          Affymetrix Rat Gene 1.0 ST Array 10765606 - -
          Affymetrix Rat Gene 1.0 ST Array 10765604 - -
          Affymetrix Rat Gene 1.0 ST Array 10765605 - -
          Affymetrix Rat Gene 1.0 ST Array 10765602 - -
          Affymetrix Rat Gene 1.0 ST Array 10765607 - -
          Affymetrix Rat Gene 1.0 ST Array 10765603 - -
          Affymetrix Rat Gene 1.1 ST Array 10765600 - -
          Rat Genome 230 2.0 Array 1387279_PM_at - -
          Rat Genome 230 2.0 Array 1387279_at - -
          Affymetrix HT Rat Focus Array 1387279_at - -
          Affymetrix HT Rat Focus Array 1387279_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300005375.rn.1 TC1300000765.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005376.rn.1 TC1300000765.rn.1 RTA TC
          Rat Expression Set 230 1387279_at - -
          Rat Expression Set 230 1387279_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10765600 - -
          Rat Genome U34 Set rc_AA818119_at - -
          Rat Genome U34 Set rc_AI232375_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21558126 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351536 - -
          RatRef-12 Expression BeadChip ILMN_69716 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline