Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587437_m1
          See other TXN1 GE Assays ›
          Gene Symbol:
          Txn1
          Gene Name:
          thioredoxin 1
          Gene Aliases:
          Txn
          Chromosome Location:
          Chr.5: 75049735 - 75057731 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587437_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Txn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053800.3 NP_446252.1 2 - 3 193 68
          GenBank mRNA AF311055.1 - 2 - 3 193 68
          BC058454.1 - 278 6886988 68
          FQ209943.1 - 275 68
          FQ216849.1 - 304 68
          FQ217822.1 - 273 68
          FQ221646.1 - 246 68
          X14878.1 - 253 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002825.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Txn1

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          glycerol ether metabolic process
          response to radiation
          response to selenium ion
          response to activity
          positive regulation of peptidyl-serine phosphorylation
          regulation of protein import into nucleus, translocation
          cellular response to drug
          response to drug
          positive regulation of DNA binding
          cell redox homeostasis
          negative regulation of protein export from nucleus
          response to axon injury
          oxidation-reduction process
          cellular response to antibiotic
          cellular response to glucose stimulus
          cellular response to hyperoxia
          response to dexamethasone
          response to thyroxine
          negative regulation of hydrogen peroxide-induced cell death
          thioredoxin-disulfide reductase activity
          protein disulfide oxidoreductase activity
          peptide disulfide oxidoreductase activity
          enzyme binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P689013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12020 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P689013 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P689013 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P689013 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P689013 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700145130 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36505 - -
          Agilent Rat cDNA Microarray (G4105A) 700145130 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10877007 - -
          Affymetrix Rat Gene 1.0 ST Array 10877010 - -
          Affymetrix Rat Gene 1.0 ST Array 10877009 - -
          Affymetrix Rat Gene 1.0 ST Array 10877008 - -
          Affymetrix Rat Gene 1.0 ST Array 10877011 - -
          Affymetrix Rat Gene 1.0 ST Array 10877012 - -
          Affymetrix Rat Gene 1.1 ST Array 10877005 - -
          Rat Genome 230 2.0 Array 1398839_at - -
          Rat Genome 230 2.0 Array 1398839_PM_at - -
          Affymetrix HT Rat Focus Array 1398839_at - -
          Affymetrix HT Rat Focus Array 1398839_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500020862.rn.1 TC0500002825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500020865.rn.1 TC0500002825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500020866.rn.1 TC0500002825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500020863.rn.1 TC0500002825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500020868.rn.1 TC0500002825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500020864.rn.1 TC0500002825.rn.1 RTA TC
          Rat Expression Set 230 1398839_PM_at - -
          Rat Expression Set 230 1398839_at - -
          Affymetrix Rat Gene 1.0 ST Array 10877005 - -
          Rat Genome U34 Set rc_AA944316_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21986484 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371958 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038784 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038783 - -
          RatRef-12 Expression BeadChip ILMN_60131 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline