Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587469_m1
          See other GRIPAP1 GE Assays ›
          Gene Symbol:
          Gripap1
          Gene Name:
          GRIP1 associated protein 1
          Gene Aliases:
          Grasp1
          Chromosome Location:
          Chr.X: 15523929 - 15553702 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587469_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gripap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053807.1 NP_446259.1 22 - 23 2092 66
          GenBank mRNA AF274057.1 - 22 - 23 2092 66
          AF274058.1 - 20 - 21 1957 66
          FQ233233.1 - 1 - 2 91 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001498.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gripap1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P511410 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P088685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129782 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49312 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P511410 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088685 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P511410 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088685 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P511410 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P088685 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P511410 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701030191 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701911304 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62072 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101143 - -
          Agilent Rat cDNA Microarray (G4105A) 701030191 - -
          Agilent Rat cDNA Microarray (G4105A) 701911304 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937041 - -
          Affymetrix Rat Gene 1.0 ST Array 10937032 - -
          Affymetrix Rat Gene 1.0 ST Array 10937027 - -
          Affymetrix Rat Gene 1.0 ST Array 10937045 - -
          Affymetrix Rat Gene 1.0 ST Array 10937044 - -
          Affymetrix Rat Gene 1.0 ST Array 10937043 - -
          Affymetrix Rat Gene 1.0 ST Array 10937028 - -
          Affymetrix Rat Gene 1.0 ST Array 10937042 - -
          Affymetrix Rat Gene 1.0 ST Array 10937036 - -
          Affymetrix Rat Gene 1.0 ST Array 10937029 - -
          Affymetrix Rat Gene 1.0 ST Array 10937037 - -
          Affymetrix Rat Gene 1.0 ST Array 10937021 - -
          Affymetrix Rat Gene 1.0 ST Array 10937033 - -
          Affymetrix Rat Gene 1.0 ST Array 10937038 - -
          Affymetrix Rat Gene 1.0 ST Array 10937025 - -
          Affymetrix Rat Gene 1.0 ST Array 10937031 - -
          Affymetrix Rat Gene 1.0 ST Array 10937022 - -
          Affymetrix Rat Gene 1.0 ST Array 10937039 - -
          Affymetrix Rat Gene 1.0 ST Array 10937040 - -
          Affymetrix Rat Gene 1.0 ST Array 10937035 - -
          Affymetrix Rat Gene 1.0 ST Array 10937046 - -
          Affymetrix Rat Gene 1.0 ST Array 10937024 - -
          Affymetrix Rat Gene 1.0 ST Array 10937047 - -
          Affymetrix Rat Gene 1.0 ST Array 10937026 - -
          Affymetrix Rat Gene 1.0 ST Array 10937034 - -
          Affymetrix Rat Gene 1.0 ST Array 10937023 - -
          Affymetrix Rat Gene 1.0 ST Array 10937030 - -
          Affymetrix Rat Gene 1.1 ST Array 10937020 - -
          Rat Genome 230 2.0 Array 1368909_PM_a_at - -
          Rat Genome 230 2.0 Array 1368909_a_at - -
          Affymetrix HT Rat Focus Array 1368909_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368909_a_at - -
          Affymetrix Rat Clariom S Assay PSR0X00009536.rn.1 TC0X00001498.rn.1 RTA TC
          Rat Expression Set 230 1368909_PM_a_at - -
          Rat Expression Set 230 1368909_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937020 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21100914 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370036 - -
          RatRef-12 Expression BeadChip ILMN_56540 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline