Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587598_m1
          See other PDK1 GE Assays ›
          Gene Symbol:
          Pdk1
          Gene Name:
          pyruvate dehydrogenase kinase, isozyme 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 58530870 - 58561493 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587598_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053826.2 NP_446278.2 9 - 10 1159 69
          XM_006234352.2 XP_006234414.1 9 - 10 931 69
          GenBank mRNA BC089783.1 - 9 - 10 1159 7321326 69
          L22294.1 - 9 - 10 1090 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000812.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdk1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose metabolic process
          protein phosphorylation
          cell surface receptor signaling pathway
          cell proliferation
          intrinsic apoptotic signaling pathway in response to oxidative stress
          regulation of acetyl-CoA biosynthetic process from pyruvate
          regulation of glucose metabolic process
          peptidyl-serine phosphorylation
          hypoxia-inducible factor-1alpha signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          pyruvate dehydrogenase (acetyl-transferring) kinase activity
          ATP binding
          protein complex binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P139718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13994 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130577 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175135 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171280 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P139718 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P139718 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P139718 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P139718 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700483258 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9909 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100419 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2946 - -
          Agilent Rat cDNA Microarray (G4105A) 700483258 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10836889 - -
          Affymetrix Rat Gene 1.0 ST Array 10836882 - -
          Affymetrix Rat Gene 1.0 ST Array 10836883 - -
          Affymetrix Rat Gene 1.0 ST Array 10836878 - -
          Affymetrix Rat Gene 1.0 ST Array 10836884 - -
          Affymetrix Rat Gene 1.0 ST Array 10836881 - -
          Affymetrix Rat Gene 1.0 ST Array 10836885 - -
          Affymetrix Rat Gene 1.0 ST Array 10836879 - -
          Affymetrix Rat Gene 1.0 ST Array 10836887 - -
          Affymetrix Rat Gene 1.0 ST Array 10836880 - -
          Affymetrix Rat Gene 1.0 ST Array 10836888 - -
          Affymetrix Rat Gene 1.0 ST Array 10836886 - -
          Affymetrix Rat Gene 1.1 ST Array 10836877 - -
          Rat Genome 230 2.0 Array 1368079_PM_at - -
          Rat Genome 230 2.0 Array 1368079_at - -
          Affymetrix HT Rat Focus Array 1368079_at - -
          Affymetrix HT Rat Focus Array 1368079_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300006190.rn.1 TC0300000812.rn.1 RTA TC
          Rat Expression Set 230 1368079_PM_at - -
          Rat Expression Set 230 1368079_at - -
          Affymetrix Rat Gene 1.0 ST Array 10836877 - -
          Rat Genome U34 Set L22294_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21249653 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363982 - -
          RatRef-12 Expression BeadChip ILMN_47996 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline