Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587634_m1
          See other NUP107 GE Assays ›
          Gene Symbol:
          Nup107
          Gene Name:
          nucleoporin 107
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 60781724 - 60825225 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587634_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nup107
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053830.2 NP_446282.2 26 - 27 2572 56
          GenBank mRNA BQ191246.1 - 2 - 3 56
          C07146.1 - 2 - 3 200 56
          CK844566.1 - 2 - 3 56
          L31840.1 - 26 - 27 2592 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002767.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nup107

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA export from nucleus
          protein transport
          nuclear pore complex assembly
          transmembrane transport
          nucleocytoplasmic transporter activity
          porin activity
          structural constituent of nuclear pore

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P619957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212785 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32127 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59351 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159362 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67553 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P619957 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P619957 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P619957 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P472791 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700767415 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69377 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82546 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18801 - -
          Agilent Rat cDNA Microarray (G4105A) 700767415 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10902597 - -
          Affymetrix Rat Gene 1.0 ST Array 10902595 - -
          Affymetrix Rat Gene 1.0 ST Array 10902602 - -
          Affymetrix Rat Gene 1.0 ST Array 10902605 - -
          Affymetrix Rat Gene 1.0 ST Array 10902586 - -
          Affymetrix Rat Gene 1.0 ST Array 10902590 - -
          Affymetrix Rat Gene 1.0 ST Array 10902585 - -
          Affymetrix Rat Gene 1.0 ST Array 10902606 - -
          Affymetrix Rat Gene 1.0 ST Array 10902587 - -
          Affymetrix Rat Gene 1.0 ST Array 10902613 - -
          Affymetrix Rat Gene 1.0 ST Array 10902594 - -
          Affymetrix Rat Gene 1.0 ST Array 10902596 - -
          Affymetrix Rat Gene 1.0 ST Array 10902589 - -
          Affymetrix Rat Gene 1.0 ST Array 10902591 - -
          Affymetrix Rat Gene 1.0 ST Array 10902592 - -
          Affymetrix Rat Gene 1.0 ST Array 10902608 - -
          Affymetrix Rat Gene 1.0 ST Array 10902588 - -
          Affymetrix Rat Gene 1.0 ST Array 10902599 - -
          Affymetrix Rat Gene 1.0 ST Array 10902612 - -
          Affymetrix Rat Gene 1.0 ST Array 10902601 - -
          Affymetrix Rat Gene 1.0 ST Array 10902604 - -
          Affymetrix Rat Gene 1.0 ST Array 10902610 - -
          Affymetrix Rat Gene 1.0 ST Array 10902600 - -
          Affymetrix Rat Gene 1.0 ST Array 10902607 - -
          Affymetrix Rat Gene 1.0 ST Array 10902603 - -
          Affymetrix Rat Gene 1.0 ST Array 10902611 - -
          Affymetrix Rat Gene 1.0 ST Array 10902598 - -
          Affymetrix Rat Gene 1.0 ST Array 10902593 - -
          Affymetrix Rat Gene 1.0 ST Array 10902609 - -
          Affymetrix Rat Gene 1.1 ST Array 10902584 - -
          Rat Genome 230 2.0 Array 1387151_PM_at - -
          Rat Genome 230 2.0 Array 1387151_at - -
          Affymetrix HT Rat Focus Array 1387151_PM_at - -
          Affymetrix HT Rat Focus Array 1387151_at - -
          Affymetrix Rat Clariom S Assay PSR0700021572.rn.1 TC0700002767.rn.1 RTA TC
          Rat Expression Set 230 1387151_PM_at - -
          Rat Expression Set 230 1387151_at - -
          Affymetrix Rat Gene 1.0 ST Array 10902584 - -
          Rat Genome U34 Set L31840_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21790658 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351339 - -
          RatRef-12 Expression BeadChip ILMN_53098 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline