Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00588067_m1
          See other SDC3 GE Assays ›
          Gene Symbol:
          Sdc3
          Gene Name:
          syndecan 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 148923098 - 148956404 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00588067_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sdc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053893.3 NP_446345.3 3 - 4 921 137
          GenBank mRNA U52825.1 - 3 - 4 924 137
          U73184.1 - 3 - 4 989 137

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001567.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sdc3

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P929767 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39900 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119951 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8644 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109948 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P929767 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109948 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P929767 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109948 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P929767 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12972 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68118 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64135 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1109 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880125 - -
          Affymetrix Rat Gene 1.0 ST Array 10880127 - -
          Affymetrix Rat Gene 1.0 ST Array 10880132 - -
          Affymetrix Rat Gene 1.0 ST Array 10880133 - -
          Affymetrix Rat Gene 1.0 ST Array 10880129 - -
          Affymetrix Rat Gene 1.0 ST Array 10880126 - -
          Affymetrix Rat Gene 1.0 ST Array 10880134 - -
          Affymetrix Rat Gene 1.0 ST Array 10880131 - -
          Affymetrix Rat Gene 1.0 ST Array 10880130 - -
          Affymetrix Rat Gene 1.0 ST Array 10880128 - -
          Affymetrix Rat Gene 1.1 ST Array 10880124 - -
          Rat Genome 230 2.0 Array 1368581_at - -
          Rat Genome 230 2.0 Array 1368581_PM_at - -
          Rat Genome 230 2.0 Array 1372180_PM_at - -
          Rat Genome 230 2.0 Array 1372180_at - -
          Rat Genome 230 2.0 Array 1368580_PM_at - -
          Rat Genome 230 2.0 Array 1368580_at - -
          Affymetrix HT Rat Focus Array 1368580_PM_at - -
          Affymetrix HT Rat Focus Array 1368581_PM_at - -
          Affymetrix HT Rat Focus Array 1368580_at - -
          Affymetrix HT Rat Focus Array 1368581_at - -
          Affymetrix Rat Clariom S Assay PSR0500011311.rn.1 TC0500001567.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011308.rn.1 TC0500001567.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011307.rn.1 TC0500001567.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011313.rn.1 TC0500001567.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011309.rn.1 TC0500001567.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011314.rn.1 TC0500001567.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011306.rn.1 TC0500001567.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011312.rn.1 TC0500001567.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011310.rn.1 TC0500001567.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011315.rn.1 TC0500001567.rn.1 RTA TC
          Rat Expression Set 230 1368581_at - -
          Rat Expression Set 230 1368580_PM_at - -
          Rat Expression Set 230 1372180_at - -
          Rat Expression Set 230 1368581_PM_at - -
          Rat Expression Set 230 1368580_at - -
          Rat Expression Set 230 1372180_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880124 - -
          Rat Genome U34 Set X63143_s_at - -
          Rat Genome U34 Set rc_AI029642_at - -
          Rat Genome U34 Set rc_AA964067_at - -
          Rat Genome U34 Set X63143_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21353282 - -
          ABI Rat Genome Survey Microarray 21860762 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354087 - -
          RatRef-12 Expression BeadChip ILMN_69620 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline