Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00588079_m1
          See other ALDH1A2 GE Assays ›
          Gene Symbol:
          Aldh1a2
          Gene Name:
          aldehyde dehydrogenase 1 family, member A2
          Gene Aliases:
          Raldh-2
          Chromosome Location:
          Chr.8: 77640234 - 77719488 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00588079_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh1a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053896.2 NP_446348.2 13 - 14 1587 114
          GenBank mRNA BC098910.1 - 13 - 14 1579 7455646 114
          EV764818.1 - 1 - 2 112
          U60063.1 - 13 - 14 1583 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001056.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aldh1a2

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          kidney development
          liver development
          regulation of endothelial cell proliferation
          retinoic acid biosynthetic process
          heart morphogenesis
          midgut development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          determination of bilateral symmetry
          anterior/posterior pattern specification
          proximal/distal pattern formation
          positive regulation of gene expression
          neural crest cell development
          morphogenesis of embryonic epithelium
          neural tube development
          pituitary gland development
          neuron differentiation
          lung development
          embryonic limb morphogenesis
          forebrain development
          hindbrain development
          pancreas development
          embryonic camera-type eye development
          response to estradiol
          response to vitamin A
          response to cytokine
          embryonic forelimb morphogenesis
          ureter maturation
          retinol metabolic process
          retinoic acid metabolic process
          retinal metabolic process
          9-cis-retinoic acid biosynthetic process
          camera-type eye development
          positive regulation of apoptotic process
          retinoic acid receptor signaling pathway
          embryonic digestive tract development
          cardiac muscle tissue development
          oxidation-reduction process
          face development
          cellular response to retinoic acid
          retinal dehydrogenase activity
          3-chloroallyl aldehyde dehydrogenase activity
          aldehyde dehydrogenase (NAD) activity
          retinal binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P317912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122474 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92388 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115167 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3796 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P317912 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P317912 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P317912 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P317912 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73901 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38058 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73961 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6341 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95212 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10911384 - -
          Affymetrix Rat Gene 1.0 ST Array 10911381 - -
          Affymetrix Rat Gene 1.0 ST Array 10911385 - -
          Affymetrix Rat Gene 1.0 ST Array 10911391 - -
          Affymetrix Rat Gene 1.0 ST Array 10911387 - -
          Affymetrix Rat Gene 1.0 ST Array 10911388 - -
          Affymetrix Rat Gene 1.0 ST Array 10911390 - -
          Affymetrix Rat Gene 1.0 ST Array 10911392 - -
          Affymetrix Rat Gene 1.0 ST Array 10911382 - -
          Affymetrix Rat Gene 1.0 ST Array 10911394 - -
          Affymetrix Rat Gene 1.0 ST Array 10911386 - -
          Affymetrix Rat Gene 1.0 ST Array 10911383 - -
          Affymetrix Rat Gene 1.0 ST Array 10911389 - -
          Affymetrix Rat Gene 1.0 ST Array 10911393 - -
          Affymetrix Rat Gene 1.1 ST Array 10911380 - -
          Rat Genome 230 2.0 Array 1368003_at - -
          Rat Genome 230 2.0 Array 1368003_PM_at - -
          Affymetrix HT Rat Focus Array 1368003_PM_at - -
          Affymetrix HT Rat Focus Array 1368003_at - -
          Affymetrix Rat Clariom S Assay PSR0800007672.rn.1 TC0800001056.rn.1 RTA TC
          Rat Expression Set 230 1368003_at - -
          Rat Expression Set 230 1368003_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10911380 - -
          Rat Genome U34 Set U60063_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22210228 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359926 - -
          RatRef-12 Expression BeadChip ILMN_68097 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline