Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00588145_m1
          See other LMNB1 GE Assays ›
          Gene Symbol:
          Lmnb1
          Gene Name:
          lamin B1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 51785111 - 51822264 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00588145_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lmnb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053905.1 NP_446357.1 1 - 2 681 77
          GenBank mRNA U72353.1 - 1 - 2 681 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000555.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lmnb1

          Gene Ontology Categories:

          Process(es)

          structural molecule activity
          phospholipase binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P075647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897122 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171481 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32678 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166866 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075647 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075647 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P075647 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P325522 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57599 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100534 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79018 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10801826 - -
          Affymetrix Rat Gene 1.0 ST Array 10801822 - -
          Affymetrix Rat Gene 1.0 ST Array 10801827 - -
          Affymetrix Rat Gene 1.0 ST Array 10801824 - -
          Affymetrix Rat Gene 1.0 ST Array 10801828 - -
          Affymetrix Rat Gene 1.0 ST Array 10801823 - -
          Affymetrix Rat Gene 1.0 ST Array 10801832 - -
          Affymetrix Rat Gene 1.0 ST Array 10801830 - -
          Affymetrix Rat Gene 1.0 ST Array 10801825 - -
          Affymetrix Rat Gene 1.0 ST Array 10801831 - -
          Affymetrix Rat Gene 1.0 ST Array 10801829 - -
          Affymetrix Rat Gene 1.1 ST Array 10801821 - -
          Rat Genome 230 2.0 Array 1373897_PM_at - -
          Rat Genome 230 2.0 Array 1368811_at - -
          Rat Genome 230 2.0 Array 1368811_PM_at - -
          Rat Genome 230 2.0 Array 1373897_at - -
          Rat Genome 230 2.0 Array 1368812_at - -
          Rat Genome 230 2.0 Array 1368812_PM_at - -
          Affymetrix HT Rat Focus Array 1373897_at - -
          Affymetrix HT Rat Focus Array 1373897_PM_at - -
          Affymetrix HT Rat Focus Array 1368811_PM_at - -
          Affymetrix HT Rat Focus Array 1368812_PM_at - -
          Affymetrix HT Rat Focus Array 1368812_at - -
          Affymetrix HT Rat Focus Array 1368811_at - -
          Affymetrix Rat Clariom S Assay PSR1800004031.rn.1 TC1800000555.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004033.rn.1 TC1800000555.rn.1 RTA TC
          Rat Expression Set 230 1373897_PM_at - -
          Rat Expression Set 230 1368811_PM_at - -
          Rat Expression Set 230 1373897_at - -
          Rat Expression Set 230 1368812_at - -
          Rat Expression Set 230 1368811_at - -
          Rat Expression Set 230 1368812_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10801821 - -
          Rat Genome U34 Set U72353_at - -
          Rat Genome U34 Set rc_AA957201_s_at - -
          Rat Genome U34 Set rc_AI103106_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21505339 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline