Search
Search
| Gene Symbol: Psmd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053925.1 | NP_446377.1 | 216 | 106 | ||
| GenBank mRNA | AB022014.1 | - | 216 | 106 | ||
| BC158584.1 | - | 226 | 8369120 | 106 | ||
Set Membership: |
|