Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00588589_m1
          See other IL1R2 GE Assays ›
          Gene Symbol:
          Il1r2
          Gene Name:
          interleukin 1 receptor, type II
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 46840642 - 46881239 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00588589_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il1r2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053953.1 NP_446405.1 2 - 3 228 66
          XM_008766983.1 XP_008765205.1 2 - 3 613 66
          XM_008766984.1 XP_008765206.1 2 - 3 384 66
          GenBank mRNA BC091564.1 - 2 - 3 202 7445971 66
          FQ228742.1 - 2 - 3 227 66
          Z22812.1 - 2 - 3 228 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000532.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il1r2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine-mediated signaling pathway
          interleukin-1, Type II, blocking receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P820657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173454 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4307 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8056 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P820657 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P820657 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P820657 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P820657 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90617 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11561 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25925 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10922819 - -
          Affymetrix Rat Gene 1.0 ST Array 10922821 - -
          Affymetrix Rat Gene 1.0 ST Array 10922817 - -
          Affymetrix Rat Gene 1.0 ST Array 10922824 - -
          Affymetrix Rat Gene 1.0 ST Array 10922822 - -
          Affymetrix Rat Gene 1.0 ST Array 10922818 - -
          Affymetrix Rat Gene 1.0 ST Array 10922820 - -
          Affymetrix Rat Gene 1.0 ST Array 10922825 - -
          Affymetrix Rat Gene 1.0 ST Array 10922823 - -
          Affymetrix Rat Gene 1.1 ST Array 10922816 - -
          Rat Genome 230 2.0 Array 1387180_PM_at - -
          Rat Genome 230 2.0 Array 1387180_at - -
          Affymetrix HT Rat Focus Array 1387180_at - -
          Affymetrix HT Rat Focus Array 1387180_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900004330.rn.1 TC0900000532.rn.1 RTA TC
          Rat Expression Set 230 1387180_PM_at - -
          Rat Expression Set 230 1387180_at - -
          Affymetrix Rat Gene 1.0 ST Array 10922816 - -
          Rat Genome U34 Set Z22812_at - -
          Rat Genome U34 Set Z22812_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22027162 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349030 - -
          RatRef-12 Expression BeadChip ILMN_49738 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline