Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053954.1 | NP_446406.1 | 3 - 4 | 1257 | 73 | |
XM_006235509.2 | XP_006235571.1 | 2 - 3 | 1673 | 73 | ||
GenBank mRNA | Y17606.1 | - | 3 - 4 | 1257 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |