Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00588832_m1
          See other UBR5 GE Assays ›
          Gene Symbol:
          Ubr5
          Gene Name:
          ubiquitin protein ligase E3 component n-recognin 5
          Gene Aliases:
          Dd5, Rat100
          Chromosome Location:
          Chr.7: 76789110 - 76835518 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00588832_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubr5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006241570.2 XP_006241632.2 3768 61
          XM_008765468.1 XP_008763690.1 3819 61
          XM_008765469.1 XP_008763691.1 3819 61
          GenBank mRNA X64411.1 - 58 61

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the X64411.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          DNA repair
          cellular response to DNA damage stimulus
          protein ubiquitination
          positive regulation of protein import into nucleus, translocation
          positive regulation of catenin import into nucleus
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          progesterone receptor signaling pathway
          positive regulation of canonical Wnt signaling pathway
          negative regulation of histone H2A K63-linked ubiquitination
          negative regulation of double-strand break repair
          RNA binding
          ubiquitin-protein transferase activity
          zinc ion binding
          ligase activity
          protein domain specific binding
          ubiquitin-ubiquitin ligase activity
          ubiquitin binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P977977 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P168132 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P168132 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P168132 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P168132 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P984629 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P977977 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P729238 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20948 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P898673 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P168132 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701335508 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700494843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102853 - -
          Agilent Rat cDNA Microarray (G4105A) 701335508 - -
          Agilent Rat cDNA Microarray (G4105A) 700494843 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903412 - -
          Affymetrix Rat Gene 1.0 ST Array 10903406 - -
          Affymetrix Rat Gene 1.0 ST Array 10903416 - -
          Affymetrix Rat Gene 1.0 ST Array 10903414 - -
          Affymetrix Rat Gene 1.0 ST Array 10903420 - -
          Affymetrix Rat Gene 1.0 ST Array 10903408 - -
          Affymetrix Rat Gene 1.0 ST Array 10903404 - -
          Affymetrix Rat Gene 1.0 ST Array 10903401 - -
          Affymetrix Rat Gene 1.0 ST Array 10903403 - -
          Affymetrix Rat Gene 1.0 ST Array 10903410 - -
          Affymetrix Rat Gene 1.0 ST Array 10903405 - -
          Affymetrix Rat Gene 1.0 ST Array 10903407 - -
          Affymetrix Rat Gene 1.0 ST Array 10903409 - -
          Affymetrix Rat Gene 1.0 ST Array 10903415 - -
          Affymetrix Rat Gene 1.0 ST Array 10903417 - -
          Affymetrix Rat Gene 1.0 ST Array 10903419 - -
          Affymetrix Rat Gene 1.0 ST Array 10903402 - -
          Affymetrix Rat Gene 1.0 ST Array 10903411 - -
          Affymetrix Rat Gene 1.0 ST Array 10903413 - -
          Affymetrix Rat Gene 1.0 ST Array 10903418 - -
          Affymetrix Rat Gene 1.1 ST Array 10903400 - -
          Rat Genome 230 2.0 Array 1392199_at - -
          Rat Genome 230 2.0 Array 1367719_at - -
          Rat Genome 230 2.0 Array 1392199_PM_at - -
          Rat Genome 230 2.0 Array 1367719_PM_at - -
          Affymetrix HT Rat Focus Array 1367719_PM_at - -
          Affymetrix HT Rat Focus Array 1367719_at - -
          Rat Expression Set 230 1367719_PM_at - -
          Rat Expression Set 230 1367719_at - -
          Rat Expression Set 230 1392199_at - -
          Rat Expression Set 230 1392199_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903400 - -
          Rat Genome U34 Set rc_AI237592_at - -
          Rat Genome U34 Set rc_AA997087_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040780 - -
          RatRef-12 Expression BeadChip ILMN_64281 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline