Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00588964_m1
          See other VAMP3 GE Assays ›
          Gene Symbol:
          Vamp3
          Gene Name:
          vesicle-associated membrane protein 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 168126157 - 168136628 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00588964_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vamp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057097.2 NP_476438.1 3 - 4 297 136
          GenBank mRNA BC088119.1 - 3 - 4 297 7366421 136
          S63830.1 - 349 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500004117.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vamp3

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of receptor recycling
          protein complex assembly
          exocytosis
          vesicle fusion
          neurotransmitter secretion
          vesicle-mediated transport
          calcium ion regulated exocytosis
          substrate adhesion-dependent cell spreading
          SNARE complex assembly
          retrograde transport, endosome to Golgi
          Golgi to plasma membrane protein transport
          positive regulation of immunoglobulin secretion
          mucus secretion
          negative regulation of secretion by cell
          regulation of histamine secretion by mast cell
          SNARE binding
          SNAP receptor activity
          protein binding
          syntaxin-1 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804636 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164993 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122653 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14574 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14574 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14574 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14574 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700306410 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62438 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103413 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59849 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7212 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1784 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87981 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64000 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55664 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2730 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70662 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77663 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84293 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35580 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70152 - -
          Agilent Rat cDNA Microarray (G4105A) 700306410 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10881888 - -
          Affymetrix Rat Gene 1.0 ST Array 10881890 - -
          Affymetrix Rat Gene 1.0 ST Array 10881889 - -
          Affymetrix Rat Gene 1.0 ST Array 10881887 - -
          Affymetrix Rat Gene 1.0 ST Array 10881886 - -
          Affymetrix Rat Gene 1.0 ST Array 10881891 - -
          Affymetrix Rat Gene 1.1 ST Array 10881885 - -
          Rat Genome 230 2.0 Array 1399159_a_at - -
          Rat Genome 230 2.0 Array 1399159_PM_a_at - -
          Affymetrix HT Rat Focus Array 1399159_PM_a_at - -
          Affymetrix HT Rat Focus Array 1399159_a_at - -
          Affymetrix Rat Clariom S Assay PSR0500031467.rn.1 TC0500004117.rn.1 RTA TC
          Rat Expression Set 230 1399159_PM_a_at - -
          Rat Expression Set 230 1399159_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10881885 - -
          Rat Genome U34 Set S63830_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21268938 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373041 - -
          RatRef-12 Expression BeadChip ILMN_61488 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline