Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589262_m1
          See other C9 GE Assays ›
          Gene Symbol:
          C9
          Gene Name:
          complement component 9
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 55775562 - 55823807 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589262_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057146.1 NP_476487.1 1 - 2 134 89
          GenBank mRNA BC091127.1 - 1 - 2 130 7324872 89
          U49071.1 - 1 - 2 116 89
          U52948.1 - 1 - 2 134 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000504.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C9

          Gene Ontology Categories:

          Function(s)

          immune response
          complement activation, alternative pathway
          complement activation, classical pathway
          blood coagulation
          cytolysis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P274130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2165 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138441 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4461 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131564 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P274130 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P274130 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P274130 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P274130 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700304375 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82844 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55591 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3090 - -
          Agilent Rat cDNA Microarray (G4105A) 700304375 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813385 - -
          Affymetrix Rat Gene 1.0 ST Array 10813387 - -
          Affymetrix Rat Gene 1.0 ST Array 10813382 - -
          Affymetrix Rat Gene 1.0 ST Array 10813381 - -
          Affymetrix Rat Gene 1.0 ST Array 10813386 - -
          Affymetrix Rat Gene 1.0 ST Array 10813388 - -
          Affymetrix Rat Gene 1.0 ST Array 10813390 - -
          Affymetrix Rat Gene 1.0 ST Array 10813383 - -
          Affymetrix Rat Gene 1.0 ST Array 10813384 - -
          Affymetrix Rat Gene 1.0 ST Array 10813389 - -
          Affymetrix Rat Gene 1.0 ST Array 10813391 - -
          Affymetrix Rat Gene 1.1 ST Array 10813380 - -
          Rat Genome 230 2.0 Array 1368741_PM_at - -
          Rat Genome 230 2.0 Array 1368741_at - -
          Affymetrix HT Rat Focus Array 1368741_at - -
          Affymetrix HT Rat Focus Array 1368741_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200002999.rn.1 TC0200000504.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200003000.rn.1 TC0200000504.rn.1 RTA TC
          Rat Expression Set 230 1368741_PM_at - -
          Rat Expression Set 230 1368741_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813380 - -
          Rat Genome U34 Set U52948_at - -
          Rat Genome U34 Set U52948_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21613623 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651040 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline