Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589273_m1
          See other SEC22A GE Assays ›
          Gene Symbol:
          Sec22a
          Gene Name:
          SEC22 vesicle trafficking protein homolog A (S. cerevisiae)
          Gene Aliases:
          Sec22l2
          Chromosome Location:
          Chr.11: 68632709 - 68686553 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589273_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sec22a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057147.2 NP_476488.2 4 - 5 577 68
          XM_006248419.2 XP_006248481.1 5 - 6 779 68
          XM_006248420.2 XP_006248482.1 4 - 5 814 68
          XM_008768692.1 XP_008766914.1 4 - 5 1026 68
          XM_008768693.1 XP_008766915.1 4 - 5 1028 68
          GenBank mRNA BC081746.1 - 4 - 5 577 7135589 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000707.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sec22a

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          ER to Golgi vesicle-mediated transport
          protein transport
          vesicle fusion with Golgi apparatus
          SNARE binding
          SNAP receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135832 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164686 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13046 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13046 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13046 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13046 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86854 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12080 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940660 - -
          Affymetrix Rat Gene 1.1 ST Array 10940660 - -
          Rat Genome 230 2.0 Array 1369092_at - -
          Rat Genome 230 2.0 Array 1369092_PM_at - -
          Affymetrix HT Rat Focus Array 1369092_at - -
          Affymetrix HT Rat Focus Array 1369092_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100003870.rn.1 TC1100000707.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003869.rn.1 TC1100000707.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003868.rn.1 TC1100000707.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003873.rn.1 TC1100000707.rn.1 RTA TC
          Rat Expression Set 230 1369092_at - -
          Rat Expression Set 230 1369092_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940660 - -
          Rat Genome U34 Set U42209_at - -
          Rat Genome U34 Set rc_AI233439_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21913868 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366022 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline