Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589313_m1
          See other XPNPEP2 GE Assays ›
          Gene Symbol:
          Xpnpep2
          Gene Name:
          X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 134940699 - 134969874 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589313_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xpnpep2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057155.3 NP_476496.1 2 - 3 163 73
          XM_008773513.1 XP_008771735.1 3 - 4 603 73
          XM_008773514.1 XP_008771736.1 2 - 3 466 73
          GenBank mRNA AF359355.1 - 2 - 3 163 73
          BC074017.1 - 2 - 3 427 7098467 73

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_008773514.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Xpnpep2

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          biological_process
          molecular_function
          aminopeptidase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P386724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530653 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92634 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79502 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P386724 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P386724 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P386724 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P386724 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53667 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30426 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10500 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10935310 - -
          Affymetrix Rat Gene 1.0 ST Array 10935306 - -
          Affymetrix Rat Gene 1.0 ST Array 10935300 - -
          Affymetrix Rat Gene 1.0 ST Array 10935298 - -
          Affymetrix Rat Gene 1.0 ST Array 10935292 - -
          Affymetrix Rat Gene 1.0 ST Array 10935294 - -
          Affymetrix Rat Gene 1.0 ST Array 10935308 - -
          Affymetrix Rat Gene 1.0 ST Array 10935313 - -
          Affymetrix Rat Gene 1.0 ST Array 10935303 - -
          Affymetrix Rat Gene 1.0 ST Array 10935304 - -
          Affymetrix Rat Gene 1.0 ST Array 10935305 - -
          Affymetrix Rat Gene 1.0 ST Array 10935290 - -
          Affymetrix Rat Gene 1.0 ST Array 10935297 - -
          Affymetrix Rat Gene 1.0 ST Array 10935291 - -
          Affymetrix Rat Gene 1.0 ST Array 10935295 - -
          Affymetrix Rat Gene 1.0 ST Array 10935296 - -
          Affymetrix Rat Gene 1.0 ST Array 10935311 - -
          Affymetrix Rat Gene 1.0 ST Array 10935301 - -
          Affymetrix Rat Gene 1.0 ST Array 10935309 - -
          Affymetrix Rat Gene 1.0 ST Array 10935302 - -
          Affymetrix Rat Gene 1.0 ST Array 10935299 - -
          Affymetrix Rat Gene 1.0 ST Array 10935307 - -
          Affymetrix Rat Gene 1.0 ST Array 10935293 - -
          Affymetrix Rat Gene 1.0 ST Array 10935312 - -
          Affymetrix Rat Gene 1.1 ST Array 10935289 - -
          Rat Genome 230 2.0 Array 1368428_at - -
          Rat Genome 230 2.0 Array 1368428_PM_at - -
          Affymetrix HT Rat Focus Array 1368428_at - -
          Affymetrix HT Rat Focus Array 1368428_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00006952.rn.1 TC0X00001140.rn.1 RTA TC
          Rat Expression Set 230 1368428_PM_at - -
          Rat Expression Set 230 1368428_at - -
          Affymetrix Rat Gene 1.0 ST Array 10935289 - -
          Rat Genome U34 Set rc_AA946466_at - -
          Rat Genome U34 Set rc_AA946466_g_at - -
          Rat Genome U34 Set rc_AA818739_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21483105 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369859 - -
          RatRef-12 Expression BeadChip ILMN_62821 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline