Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589333_m1
          See other FOLH1 GE Assays ›
          Gene Symbol:
          Folh1
          Gene Name:
          folate hydrolase 1
          Gene Aliases:
          Naalad
          Chromosome Location:
          Chr.1: 150323768 - 150395415 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589333_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Folh1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057185.2 NP_476533.1 13 - 14 1514 129
          GenBank mRNA AF039707.1 - 7 - 8 600 129
          AF040256.1 - 13 - 14 1520 129
          AF513486.2 - 13 - 14 1514 129
          U75973.1 - 13 - 14 1471 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006006.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Folh1

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          folic acid-containing compound metabolic process
          carboxypeptidase activity
          peptidase activity
          metallopeptidase activity
          dipeptidase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P159767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618594 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64637 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39464 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102066 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85948 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25353 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P159767 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P159767 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P159767 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P657469 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103608 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42563 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51985 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44603 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21340 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10723528 - -
          Affymetrix Rat Gene 1.0 ST Array 10723544 - -
          Affymetrix Rat Gene 1.0 ST Array 10723530 - -
          Affymetrix Rat Gene 1.0 ST Array 10723533 - -
          Affymetrix Rat Gene 1.0 ST Array 10723529 - -
          Affymetrix Rat Gene 1.0 ST Array 10723526 - -
          Affymetrix Rat Gene 1.0 ST Array 10723536 - -
          Affymetrix Rat Gene 1.0 ST Array 10723534 - -
          Affymetrix Rat Gene 1.0 ST Array 10723537 - -
          Affymetrix Rat Gene 1.0 ST Array 10723543 - -
          Affymetrix Rat Gene 1.0 ST Array 10723524 - -
          Affymetrix Rat Gene 1.0 ST Array 10723527 - -
          Affymetrix Rat Gene 1.0 ST Array 10723539 - -
          Affymetrix Rat Gene 1.0 ST Array 10723525 - -
          Affymetrix Rat Gene 1.0 ST Array 10723532 - -
          Affymetrix Rat Gene 1.0 ST Array 10723538 - -
          Affymetrix Rat Gene 1.0 ST Array 10723531 - -
          Affymetrix Rat Gene 1.0 ST Array 10723542 - -
          Affymetrix Rat Gene 1.0 ST Array 10723535 - -
          Affymetrix Rat Gene 1.0 ST Array 10723540 - -
          Affymetrix Rat Gene 1.0 ST Array 10723541 - -
          Affymetrix Rat Gene 1.1 ST Array 10723523 - -
          Rat Genome 230 2.0 Array 1387363_PM_at - -
          Rat Genome 230 2.0 Array 1387364_at - -
          Rat Genome 230 2.0 Array 1387364_PM_at - -
          Rat Genome 230 2.0 Array 1387363_at - -
          Affymetrix HT Rat Focus Array 1387363_PM_at - -
          Affymetrix HT Rat Focus Array 1387363_at - -
          Affymetrix HT Rat Focus Array 1387364_PM_at - -
          Affymetrix HT Rat Focus Array 1387364_at - -
          Affymetrix Rat Clariom S Assay PSR0100047758.rn.1 TC0100006006.rn.1 RTA TC
          Rat Expression Set 230 1387363_at - -
          Rat Expression Set 230 1387363_PM_at - -
          Rat Expression Set 230 1387364_PM_at - -
          Rat Expression Set 230 1387364_at - -
          Affymetrix Rat Gene 1.0 ST Array 10723523 - -
          Rat Genome U34 Set U75973_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22098122 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362507 - -
          RatRef-12 Expression BeadChip ILMN_48313 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline