Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589352_m1
          See other HADH GE Assays ›
          Gene Symbol:
          Hadh
          Gene Name:
          hydroxyacyl-CoA dehydrogenase
          Gene Aliases:
          Hadhsc
          Chromosome Location:
          Chr.2: 236353445 - 236395067 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589352_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hadh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057186.2 NP_476534.1 6 - 7 736 72
          GenBank mRNA AF095449.1 - 6 - 7 736 72
          FQ212781.1 - 6 - 7 716 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004703.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hadh

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid beta-oxidation
          response to hormone
          response to activity
          response to insulin
          response to drug
          negative regulation of insulin secretion
          oxidation-reduction process
          3-hydroxyacyl-CoA dehydrogenase activity
          NAD+ binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316656 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176706 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71587 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112416 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132756 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13054 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13054 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13054 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13054 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P528621 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700541032 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15526 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1329 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58259 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33079 - -
          Agilent Rat cDNA Microarray (G4105A) 700541032 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10826833 - -
          Affymetrix Rat Gene 1.0 ST Array 10826830 - -
          Affymetrix Rat Gene 1.0 ST Array 10826829 - -
          Affymetrix Rat Gene 1.0 ST Array 10826836 - -
          Affymetrix Rat Gene 1.0 ST Array 10826832 - -
          Affymetrix Rat Gene 1.0 ST Array 10826834 - -
          Affymetrix Rat Gene 1.0 ST Array 10826835 - -
          Affymetrix Rat Gene 1.0 ST Array 10826831 - -
          Affymetrix Rat Gene 1.1 ST Array 10826828 - -
          Rat Genome 230 2.0 Array 1389922_at - -
          Rat Genome 230 2.0 Array 1370237_at - -
          Rat Genome 230 2.0 Array 1370237_PM_at - -
          Rat Genome 230 2.0 Array 1389922_PM_at - -
          Affymetrix HT Rat Focus Array 1389922_PM_at - -
          Affymetrix HT Rat Focus Array 1370237_PM_at - -
          Affymetrix HT Rat Focus Array 1389922_at - -
          Affymetrix HT Rat Focus Array 1370237_at - -
          Affymetrix Rat Clariom S Assay PSR0200032713.rn.1 TC0200004703.rn.1 RTA TC
          Rat Expression Set 230 1389922_PM_at - -
          Rat Expression Set 230 1370237_PM_at - -
          Rat Expression Set 230 1389922_at - -
          Rat Expression Set 230 1370237_at - -
          Affymetrix Rat Gene 1.0 ST Array 10826828 - -
          Rat Genome U34 Set rc_AA891362_at - -
          Rat Genome U34 Set rc_AA848758_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21211004 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366070 - -
          RatRef-12 Expression BeadChip ILMN_60853 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline