Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589383_m1
          See other WIPF1 GE Assays ›
          Gene Symbol:
          Wipf1
          Gene Name:
          WAS/WASL interacting protein family, member 1
          Gene Aliases:
          Waspip, Wip
          Chromosome Location:
          Chr.3: 60150001 - 60207125 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589383_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wipf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057192.2 NP_476540.2 6 - 7 1569 60
          GenBank mRNA AJ303456.1 - 5 - 6 1402 60
          BC072475.1 - 6 - 7 1569 7100751 60

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wipf1

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          actin polymerization or depolymerization
          actin cytoskeleton organization
          actin filament-based movement
          positive regulation of protein export from nucleus
          response to other organism
          actin binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P200463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895599 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100965 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5927 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P200463 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P200463 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P200463 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P200463 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36212 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11332 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62802 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8023 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10846204 - -
          Affymetrix Rat Gene 1.0 ST Array 10846203 - -
          Affymetrix Rat Gene 1.0 ST Array 10846209 - -
          Affymetrix Rat Gene 1.0 ST Array 10846206 - -
          Affymetrix Rat Gene 1.0 ST Array 10846207 - -
          Affymetrix Rat Gene 1.0 ST Array 10846202 - -
          Affymetrix Rat Gene 1.0 ST Array 10846205 - -
          Affymetrix Rat Gene 1.0 ST Array 10846208 - -
          Affymetrix Rat Gene 1.1 ST Array 10846201 - -
          Rat Genome 230 2.0 Array 1382283_at - -
          Rat Genome 230 2.0 Array 1387227_PM_at - -
          Rat Genome 230 2.0 Array 1387227_at - -
          Rat Genome 230 2.0 Array 1382283_PM_at - -
          Affymetrix HT Rat Focus Array 1382283_at - -
          Affymetrix HT Rat Focus Array 1382283_PM_at - -
          Affymetrix HT Rat Focus Array 1387227_at - -
          Affymetrix HT Rat Focus Array 1387227_PM_at - -
          Rat Expression Set 230 1387227_PM_at - -
          Rat Expression Set 230 1387227_at - -
          Rat Expression Set 230 1382283_PM_at - -
          Rat Expression Set 230 1382283_at - -
          Affymetrix Rat Gene 1.0 ST Array 10846201 - -
          Rat Genome U34 Set rc_AA924338_at - -
          Rat Genome U34 Set rc_AI008158_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22378251 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362680 - -
          RatRef-12 Expression BeadChip ILMN_49311 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline