Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589411_m1
          See other BAIAP2 GE Assays ›
          Gene Symbol:
          Baiap2
          Gene Name:
          BAI1-associated protein 2
          Gene Aliases:
          Irsp53
          Chromosome Location:
          Chr.10: 109107326 - 109187458 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589411_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Baiap2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057196.1 NP_476544.1 3 - 4 278 97
          XM_006247869.2 XP_006247931.1 3 - 4 341 97
          XM_006247870.2 XP_006247932.1 3 - 4 341 97
          XM_006247871.2 XP_006247933.1 3 - 4 341 97
          XM_006247872.2 XP_006247934.1 3 - 4 341 97
          XM_008768464.1 XP_008766686.1 3 - 4 341 97
          GenBank mRNA AY037934.1 - 3 - 4 278 97
          BC074009.1 - 3 - 4 312 7099884 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002058.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Baiap2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          plasma membrane organization
          regulation of cell shape
          response to bacterium
          dendrite development
          regulation of actin cytoskeleton organization
          regulation of synaptic plasticity
          actin filament bundle assembly
          actin crosslink formation
          positive regulation of dendritic spine morphogenesis
          protein binding
          cytoskeletal adaptor activity
          PDZ domain binding
          identical protein binding
          proline-rich region binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36444 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145291 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163917 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21324 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57958 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45274 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77218 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13057 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13057 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13057 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13057 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P128229 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101348 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10874 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97325 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54371 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10740102 - -
          Affymetrix Rat Gene 1.0 ST Array 10740104 - -
          Affymetrix Rat Gene 1.0 ST Array 10740107 - -
          Affymetrix Rat Gene 1.0 ST Array 10740105 - -
          Affymetrix Rat Gene 1.0 ST Array 10740110 - -
          Affymetrix Rat Gene 1.0 ST Array 10740103 - -
          Affymetrix Rat Gene 1.0 ST Array 10740101 - -
          Affymetrix Rat Gene 1.0 ST Array 10740096 - -
          Affymetrix Rat Gene 1.0 ST Array 10740113 - -
          Affymetrix Rat Gene 1.0 ST Array 10740097 - -
          Affymetrix Rat Gene 1.0 ST Array 10740114 - -
          Affymetrix Rat Gene 1.0 ST Array 10740095 - -
          Affymetrix Rat Gene 1.0 ST Array 10740115 - -
          Affymetrix Rat Gene 1.0 ST Array 10740099 - -
          Affymetrix Rat Gene 1.0 ST Array 10740112 - -
          Affymetrix Rat Gene 1.0 ST Array 10740098 - -
          Affymetrix Rat Gene 1.0 ST Array 10740111 - -
          Affymetrix Rat Gene 1.0 ST Array 10740106 - -
          Affymetrix Rat Gene 1.0 ST Array 10740100 - -
          Affymetrix Rat Gene 1.0 ST Array 10740109 - -
          Affymetrix Rat Gene 1.1 ST Array 10740094 - -
          Rat Genome 230 2.0 Array 1370074_PM_at - -
          Rat Genome 230 2.0 Array 1374117_PM_at - -
          Rat Genome 230 2.0 Array 1373081_at - -
          Rat Genome 230 2.0 Array 1370074_at - -
          Rat Genome 230 2.0 Array 1373081_PM_at - -
          Rat Genome 230 2.0 Array 1374117_at - -
          Affymetrix HT Rat Focus Array 1370074_at - -
          Affymetrix HT Rat Focus Array 1370074_PM_at - -
          Affymetrix HT Rat Focus Array 1374117_PM_at - -
          Affymetrix HT Rat Focus Array 1374117_at - -
          Affymetrix Rat Clariom S Assay PSR1000018611.rn.1 TC1000002058.rn.1 RTA TC
          Rat Expression Set 230 1373081_at - -
          Rat Expression Set 230 1374117_PM_at - -
          Rat Expression Set 230 1373081_PM_at - -
          Rat Expression Set 230 1370074_at - -
          Rat Expression Set 230 1374117_at - -
          Rat Expression Set 230 1370074_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10740094 - -
          Rat Genome U34 Set rc_AA998152_at - -
          Rat Genome U34 Set rc_AA819554_at - -
          Rat Genome U34 Set rc_AI228657_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20956062 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365343 - -
          RatRef-12 Expression BeadChip ILMN_64940 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline