Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589420_m1
          See other DECR1 GE Assays ›
          Gene Symbol:
          Decr1
          Gene Name:
          2,4-dienoyl CoA reductase 1, mitochondrial
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 29573893 - 29601731 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589420_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Decr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057197.2 NP_476545.2 4 - 5 465 136
          GenBank mRNA BC059120.1 - 4 - 5 453 6920489 136
          CO385941.1 - 4 - 5 450 136
          D00569.1 - 4 - 5 438 136
          FQ209689.1 - 4 - 5 454 136
          FQ210607.1 - 4 - 5 446 136
          FQ210721.1 - 4 - 5 447 136
          FQ212507.1 - 4 - 5 443 136
          FQ218387.1 - 4 - 5 443 136
          FQ219629.1 - 4 - 5 446 136
          FQ219666.1 - 4 - 5 443 136
          FQ220227.1 - 4 - 5 446 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002357.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Decr1

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid beta-oxidation
          protein homotetramerization
          2,4-dienoyl-CoA reductase (NADPH) activity
          NADPH binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P526030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101986 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60815 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64755 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P526030 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P526030 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P526030 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P526030 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700628567 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46085 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84348 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5663 - -
          Agilent Rat cDNA Microarray (G4105A) 700628567 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10875639 - -
          Affymetrix Rat Gene 1.0 ST Array 10875635 - -
          Affymetrix Rat Gene 1.0 ST Array 10875632 - -
          Affymetrix Rat Gene 1.0 ST Array 10875636 - -
          Affymetrix Rat Gene 1.0 ST Array 10875633 - -
          Affymetrix Rat Gene 1.0 ST Array 10875641 - -
          Affymetrix Rat Gene 1.0 ST Array 10875637 - -
          Affymetrix Rat Gene 1.0 ST Array 10875638 - -
          Affymetrix Rat Gene 1.0 ST Array 10875640 - -
          Affymetrix Rat Gene 1.0 ST Array 10875634 - -
          Affymetrix Rat Gene 1.1 ST Array 10875631 - -
          Rat Genome 230 2.0 Array 1367777_at - -
          Rat Genome 230 2.0 Array 1367777_PM_at - -
          Affymetrix HT Rat Focus Array 1367777_at - -
          Affymetrix HT Rat Focus Array 1367777_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500017776.rn.1 TC0500002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017783.rn.1 TC0500002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017775.rn.1 TC0500002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017782.rn.1 TC0500002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017777.rn.1 TC0500002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017774.rn.1 TC0500002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017778.rn.1 TC0500002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017781.rn.1 TC0500002357.rn.1 RTA TC
          Rat Expression Set 230 1367777_at - -
          Rat Expression Set 230 1367777_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10875631 - -
          Rat Genome U34 Set D00569_at - -
          Rat Genome U34 Set D00569_g_at - -
          Rat Genome U34 Set D00569_g_at - -
          Rat Genome U34 Set D00569_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22130436 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370450 - -
          RatRef-12 Expression BeadChip ILMN_52668 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline