Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589498_m1
          See other KLF9 GE Assays ›
          Gene Symbol:
          Klf9
          Gene Name:
          Kruppel-like factor 9
          Gene Aliases:
          Bteb, Bteb1
          Chromosome Location:
          Chr.1: 240908483 - 240933198 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589498_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klf9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057211.1 NP_476559.1 1 - 2 1017 91
          GenBank mRNA D12769.1 - 1 - 2 1017 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003374.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klf9

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          embryo implantation
          circadian rhythm
          negative regulation of keratinocyte proliferation
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          progesterone receptor signaling pathway
          cellular response to cortisol stimulus
          cellular response to thyroid hormone stimulus
          cellular response to peptide
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704641 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152914 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82134 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13064 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13064 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13064 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13064 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P322590 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701532082 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700297431 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86861 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90633 - -
          Agilent Rat cDNA Microarray (G4105A) 700297431 - -
          Agilent Rat cDNA Microarray (G4105A) 701745761 - -
          Agilent Rat cDNA Microarray (G4105A) 701532082 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10729349 - -
          Affymetrix Rat Gene 1.0 ST Array 10714415 - -
          Affymetrix Rat Gene 1.0 ST Array 10714414 - -
          Affymetrix Rat Gene 1.1 ST Array 10729348 - -
          Affymetrix Rat Gene 1.1 ST Array 10714413 - -
          Rat Genome 230 2.0 Array 1390076_PM_at - -
          Rat Genome 230 2.0 Array 1370209_PM_at - -
          Rat Genome 230 2.0 Array 1390076_at - -
          Rat Genome 230 2.0 Array 1387882_PM_at - -
          Rat Genome 230 2.0 Array 1387882_at - -
          Rat Genome 230 2.0 Array 1370209_at - -
          Affymetrix HT Rat Focus Array 1370209_at - -
          Affymetrix HT Rat Focus Array 1387882_at - -
          Affymetrix HT Rat Focus Array 1370209_PM_at - -
          Affymetrix HT Rat Focus Array 1387882_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100027462.rn.1 TC0100003374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100027461.rn.1 TC0100003374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100027458.rn.1 TC0100003374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100027460.rn.1 TC0100003374.rn.1 RTA TC
          Rat Expression Set 230 1370209_at - -
          Rat Expression Set 230 1387882_PM_at - -
          Rat Expression Set 230 1387882_at - -
          Rat Expression Set 230 1370209_PM_at - -
          Rat Expression Set 230 1390076_PM_at - -
          Rat Expression Set 230 1390076_at - -
          Affymetrix Rat Gene 1.0 ST Array 10714413 - -
          Rat Genome U34 Set D12769_g_at - -
          Rat Genome U34 Set D12769_at - -
          Rat Genome U34 Set rc_AI171973_at - -
          Rat Genome U34 Set rc_AI071565_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21045196 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366897 - -
          RatRef-12 Expression BeadChip ILMN_54308 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline