Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589573_m1
          See other SLC8A2 GE Assays ›
          Gene Symbol:
          Slc8a2
          Gene Name:
          solute carrier family 8 (sodium/calcium exchanger), member 2
          Gene Aliases:
          Ncx2
          Chromosome Location:
          Chr.1: 78029555 - 78054042 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589573_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc8a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_078619.1 NP_511174.1 7 - 8 2110 78
          GenBank mRNA U08141.1 - 7 - 8 2110 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000923.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc8a2

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          calcium ion transport
          cell communication
          telencephalon development
          response to vitamin D
          sodium ion transmembrane transport
          calcium ion transport into cytosol
          calcium ion export from cell
          calcium ion import into cell
          calcium:sodium antiporter activity
          protein binding
          calmodulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P125019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940893 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65463 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157409 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49118 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125019 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125019 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P125019 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P355902 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700785026 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97510 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59828 - -
          Agilent Rat cDNA Microarray (G4105A) 700785026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704440 - -
          Affymetrix Rat Gene 1.0 ST Array 10704442 - -
          Affymetrix Rat Gene 1.0 ST Array 10704443 - -
          Affymetrix Rat Gene 1.0 ST Array 10704447 - -
          Affymetrix Rat Gene 1.0 ST Array 10704444 - -
          Affymetrix Rat Gene 1.0 ST Array 10704441 - -
          Affymetrix Rat Gene 1.0 ST Array 10704448 - -
          Affymetrix Rat Gene 1.0 ST Array 10704449 - -
          Affymetrix Rat Gene 1.0 ST Array 10704445 - -
          Affymetrix Rat Gene 1.0 ST Array 10704438 - -
          Affymetrix Rat Gene 1.0 ST Array 10704439 - -
          Affymetrix Rat Gene 1.1 ST Array 10704437 - -
          Rat Genome 230 2.0 Array 1387564_at - -
          Rat Genome 230 2.0 Array 1387564_PM_at - -
          Affymetrix HT Rat Focus Array 1387564_PM_at - -
          Affymetrix HT Rat Focus Array 1387564_at - -
          Affymetrix Rat Clariom S Assay PSR0100006317.rn.1 TC0100000923.rn.1 RTA TC
          Rat Expression Set 230 1387564_PM_at - -
          Rat Expression Set 230 1387564_at - -
          Affymetrix Rat Gene 1.0 ST Array 10704437 - -
          Rat Genome U34 Set U08141_at - -
          Rat Genome U34 Set U08141_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22357383 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650279 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline