Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589656_m1
          See other CHST10 GE Assays ›
          Gene Symbol:
          Chst10
          Gene Name:
          carbohydrate sulfotransferase 10
          Gene Aliases:
          Hnk-1st
          Chromosome Location:
          Chr.9: 45528928 - 45559184 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589656_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chst10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080397.1 NP_536322.1 3 - 4 448 75
          XM_006244730.2 XP_006244792.1 4 - 5 597 75
          XM_006244732.2 XP_006244794.1 3 - 4 255 75
          XM_008766985.1 XP_008765207.1 3 - 4 379 75
          XM_008766987.1 XP_008765209.1 3 - 4 349 75
          GenBank mRNA AF022729.1 - 3 - 4 448 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001872.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chst10

          Panther Classification:

          Molecular Function -

          transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          learning
          long-term memory
          carbohydrate biosynthetic process
          sulfotransferase activity
          HNK-1 sulfotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P510135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929118 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20747 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89482 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69505 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33306 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P510135 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P510135 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P510135 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P510135 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52829 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65145 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10927607 - -
          Affymetrix Rat Gene 1.0 ST Array 10927609 - -
          Affymetrix Rat Gene 1.0 ST Array 10927606 - -
          Affymetrix Rat Gene 1.0 ST Array 10927608 - -
          Affymetrix Rat Gene 1.0 ST Array 10927605 - -
          Affymetrix Rat Gene 1.0 ST Array 10927604 - -
          Affymetrix Rat Gene 1.1 ST Array 10927603 - -
          Rat Genome 230 2.0 Array 1367944_at - -
          Rat Genome 230 2.0 Array 1367944_PM_at - -
          Affymetrix HT Rat Focus Array 1367944_at - -
          Affymetrix HT Rat Focus Array 1367944_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900015424.rn.1 TC0900001872.rn.1 RTA TC
          Rat Expression Set 230 1367944_at - -
          Rat Expression Set 230 1367944_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10927603 - -
          Rat Genome U34 Set AF022729_at - -
          Rat Genome U34 Set AF022729_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21854066 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368596 - -
          RatRef-12 Expression BeadChip ILMN_65556 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0