Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589714_m1
          See other MAGED2 GE Assays ›
          Gene Symbol:
          Maged2
          Gene Name:
          melanoma antigen, family D, 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 33174539 - 33182715 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589714_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Maged2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271109.1 NP_001258038.1 3 - 4 607 67
          NM_001271110.1 NP_001258039.1 3 - 4 635 67
          NM_080479.2 NP_536727.2 3 - 4 607 67
          XM_006237924.2 XP_006237986.1 3 - 4 697 67
          GenBank mRNA AJ293617.1 - 2 - 3 548 67
          AY724507.1 - 3 - 4 620 67
          BC107467.1 - 3 - 4 607 7459969 67
          FQ229095.1 - 3 - 4 620 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002376.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Maged2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P308718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600461 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P308718 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P308718 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P308718 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P308718 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P308724 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81560 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937537 - -
          Affymetrix Rat Gene 1.0 ST Array 10937534 - -
          Affymetrix Rat Gene 1.0 ST Array 10937535 - -
          Affymetrix Rat Gene 1.0 ST Array 10937530 - -
          Affymetrix Rat Gene 1.0 ST Array 10937529 - -
          Affymetrix Rat Gene 1.0 ST Array 10937531 - -
          Affymetrix Rat Gene 1.0 ST Array 10937532 - -
          Affymetrix Rat Gene 1.0 ST Array 10937526 - -
          Affymetrix Rat Gene 1.0 ST Array 10937539 - -
          Affymetrix Rat Gene 1.0 ST Array 10937528 - -
          Affymetrix Rat Gene 1.0 ST Array 10937536 - -
          Affymetrix Rat Gene 1.0 ST Array 10937527 - -
          Affymetrix Rat Gene 1.0 ST Array 10937533 - -
          Affymetrix Rat Gene 1.1 ST Array 10937525 - -
          Rat Genome 230 2.0 Array 1367744_at - -
          Rat Genome 230 2.0 Array 1367744_PM_at - -
          Affymetrix HT Rat Focus Array 1367744_PM_at - -
          Affymetrix HT Rat Focus Array 1367744_at - -
          Affymetrix Rat Clariom S Assay PSR0500017894.rn.1 TC0500002376.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017896.rn.1 TC0500002376.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017897.rn.1 TC0500002376.rn.1 RTA TC
          Rat Expression Set 230 1367744_PM_at - -
          Rat Expression Set 230 1367744_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937525 - -
          Rat Genome U34 Set rc_AI013149_at - -
          Rat Genome U34 Set rc_AA965028_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20925135 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370696 - -
          RatRef-12 Expression BeadChip ILMN_69297 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline