Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589998_m1
          See other GALE GE Assays ›
          Gene Symbol:
          Gale
          Gene Name:
          UDP-galactose-4-epimerase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 154310453 - 154314959 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589998_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gale
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080783.2 NP_542961.2 3 - 4 264 67
          XM_006239134.2 XP_006239196.1 3 - 4 1229 67
          XM_008764197.1 XP_008762419.1 3 - 4 495 67
          GenBank mRNA BC097293.1 - 3 - 4 264 7443707 67
          X53949.1 - 2 - 3 245 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001693.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gale

          Panther Classification:

          Molecular Function -

          epimerase/racemase

          Gene Ontology Categories:

          Function(s) Process(es)

          galactose metabolic process
          galactose catabolic process
          UDP-N-acetylglucosamine 4-epimerase activity
          UDP-glucose 4-epimerase activity
          protein homodimerization activity
          coenzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P267117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819242 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P267117 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P267117 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P267117 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P267117 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700519605 - -
          Agilent Rat cDNA Microarray (G4105A) 700519605 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872962 - -
          Affymetrix Rat Gene 1.0 ST Array 10872965 - -
          Affymetrix Rat Gene 1.0 ST Array 10872968 - -
          Affymetrix Rat Gene 1.0 ST Array 10872966 - -
          Affymetrix Rat Gene 1.0 ST Array 10872963 - -
          Affymetrix Rat Gene 1.0 ST Array 10872971 - -
          Affymetrix Rat Gene 1.0 ST Array 10872969 - -
          Affymetrix Rat Gene 1.0 ST Array 10872960 - -
          Affymetrix Rat Gene 1.0 ST Array 10872961 - -
          Affymetrix Rat Gene 1.0 ST Array 10872967 - -
          Affymetrix Rat Gene 1.0 ST Array 10872970 - -
          Affymetrix Rat Gene 1.0 ST Array 10872964 - -
          Affymetrix Rat Gene 1.1 ST Array 10872959 - -
          Rat Genome 230 2.0 Array 1368074_PM_at - -
          Rat Genome 230 2.0 Array 1368074_at - -
          Affymetrix HT Rat Focus Array 1368074_at - -
          Affymetrix HT Rat Focus Array 1368074_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500012317.rn.1 TC0500001693.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012316.rn.1 TC0500001693.rn.1 RTA TC
          Rat Expression Set 230 1368074_at - -
          Rat Expression Set 230 1368074_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872959 - -
          Rat Genome U34 Set X53949cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21378909 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356000 - -
          RatRef-12 Expression BeadChip ILMN_62991 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline