Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590028_m1
          See other DGKA GE Assays ›
          Gene Symbol:
          Dgka
          Gene Name:
          diacylglycerol kinase, alpha
          Gene Aliases:
          Dagk1
          Chromosome Location:
          Chr.7: 3143826 - 3170768 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590028_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dgka
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080787.1 NP_542965.1 17 - 18 1935 105
          XM_008765012.1 XP_008763234.1 16 - 17 2937 105
          XM_008765013.1 XP_008763235.1 16 - 17 2937 105
          XM_008765014.1 XP_008763236.1 16 - 17 2847 105
          XM_008765015.1 XP_008763237.1 16 - 17 2779 105
          XM_008765016.1 XP_008763238.1 16 - 17 2020 105
          XM_008765017.1 XP_008763239.1 17 - 18 2805 105
          XM_008765018.1 XP_008763240.1 17 - 18 2715 105
          XM_008765019.1 XP_008763241.1 17 - 18 2714 105
          XM_008765020.1 XP_008763242.1 17 - 18 2641 105
          GenBank mRNA FQ209812.1 - 17 - 18 1829 105
          S49760.1 - 17 - 18 1935 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002005.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dgka

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein kinase C-activating G-protein coupled receptor signaling pathway
          phosphorylation
          intracellular signal transduction
          NAD+ kinase activity
          diacylglycerol kinase activity
          calcium ion binding
          ATP binding
          phospholipid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P461505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166288 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71532 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36283 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P461505 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P461505 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P461505 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P461505 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700330587 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73002 - -
          Agilent Rat cDNA Microarray (G4105A) 700330587 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10899915 - -
          Affymetrix Rat Gene 1.0 ST Array 10899937 - -
          Affymetrix Rat Gene 1.0 ST Array 10899936 - -
          Affymetrix Rat Gene 1.0 ST Array 10899922 - -
          Affymetrix Rat Gene 1.0 ST Array 10899927 - -
          Affymetrix Rat Gene 1.0 ST Array 10899929 - -
          Affymetrix Rat Gene 1.0 ST Array 10899924 - -
          Affymetrix Rat Gene 1.0 ST Array 10899914 - -
          Affymetrix Rat Gene 1.0 ST Array 10899928 - -
          Affymetrix Rat Gene 1.0 ST Array 10899933 - -
          Affymetrix Rat Gene 1.0 ST Array 10899932 - -
          Affymetrix Rat Gene 1.0 ST Array 10899916 - -
          Affymetrix Rat Gene 1.0 ST Array 10899925 - -
          Affymetrix Rat Gene 1.0 ST Array 10899920 - -
          Affymetrix Rat Gene 1.0 ST Array 10899923 - -
          Affymetrix Rat Gene 1.0 ST Array 10899917 - -
          Affymetrix Rat Gene 1.0 ST Array 10899934 - -
          Affymetrix Rat Gene 1.0 ST Array 10899935 - -
          Affymetrix Rat Gene 1.0 ST Array 10899930 - -
          Affymetrix Rat Gene 1.0 ST Array 10899918 - -
          Affymetrix Rat Gene 1.0 ST Array 10899919 - -
          Affymetrix Rat Gene 1.0 ST Array 10899926 - -
          Affymetrix Rat Gene 1.0 ST Array 10899931 - -
          Affymetrix Rat Gene 1.0 ST Array 10899921 - -
          Affymetrix Rat Gene 1.1 ST Array 10899913 - -
          Rat Genome 230 2.0 Array 1387190_at - -
          Rat Genome 230 2.0 Array 1387190_PM_at - -
          Affymetrix HT Rat Focus Array 1387190_at - -
          Affymetrix HT Rat Focus Array 1387190_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700015822.rn.1 TC0700002005.rn.1 RTA TC
          Rat Expression Set 230 1387190_PM_at - -
          Rat Expression Set 230 1387190_at - -
          Affymetrix Rat Gene 1.0 ST Array 10899913 - -
          Rat Genome U34 Set S49760_at - -
          Rat Genome U34 Set S49760_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21443918 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039333 - -
          RatRef-12 Expression BeadChip ILMN_64139 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline