Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590076_m1
          See other AS3MT GE Assays ›
          Gene Symbol:
          As3mt
          Gene Name:
          arsenite methyltransferase
          Gene Aliases:
          Cyt19
          Chromosome Location:
          Chr.1: 266482858 - 266514570 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590076_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: As3mt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080890.1 NP_543166.1 6 - 7 570 67
          GenBank mRNA AF393243.1 - 6 - 7 570 67
          BC061533.1 - 6 - 7 594 6918024 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003752.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene As3mt

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein methylation
          toxin metabolic process
          arsonoacetate metabolic process
          methylation
          response to arsenic-containing substance
          protein methyltransferase activity
          S-adenosylmethionine-dependent methyltransferase activity
          arsenite methyltransferase activity
          methylarsonite methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586878 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101878 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7286 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133461 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13091 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13091 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13091 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P372419 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13091 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95679 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63916 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51529 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715879 - -
          Affymetrix Rat Gene 1.0 ST Array 10715884 - -
          Affymetrix Rat Gene 1.0 ST Array 10715887 - -
          Affymetrix Rat Gene 1.0 ST Array 10715876 - -
          Affymetrix Rat Gene 1.0 ST Array 10715880 - -
          Affymetrix Rat Gene 1.0 ST Array 10715877 - -
          Affymetrix Rat Gene 1.0 ST Array 10715886 - -
          Affymetrix Rat Gene 1.0 ST Array 10715888 - -
          Affymetrix Rat Gene 1.0 ST Array 10715885 - -
          Affymetrix Rat Gene 1.0 ST Array 10715882 - -
          Affymetrix Rat Gene 1.0 ST Array 10715883 - -
          Affymetrix Rat Gene 1.0 ST Array 10715878 - -
          Affymetrix Rat Gene 1.0 ST Array 10715881 - -
          Affymetrix Rat Gene 1.1 ST Array 10715875 - -
          Rat Genome 230 2.0 Array 1368244_at - -
          Rat Genome 230 2.0 Array 1368244_PM_at - -
          Affymetrix HT Rat Focus Array 1368244_at - -
          Affymetrix HT Rat Focus Array 1368244_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100030659.rn.1 TC0100003752.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100030660.rn.1 TC0100003752.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100030661.rn.1 TC0100003752.rn.1 RTA TC
          Rat Expression Set 230 1368244_at - -
          Rat Expression Set 230 1368244_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10715875 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21640156 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349101 - -
          RatRef-12 Expression BeadChip ILMN_50501 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline