Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590117_m1
          See other PDE7B GE Assays ›
          Gene Symbol:
          Pde7b
          Gene Name:
          phosphodiesterase 7B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 15889845 - 16203909 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590117_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pde7b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080894.2 NP_543170.1 7 - 8 680 64
          XM_008758591.1 XP_008756813.1 6 - 7 1000 64
          XM_008758592.1 XP_008756814.1 6 - 7 584 64
          GenBank mRNA AB057409.1 - 7 - 8 680 64
          AB057410.1 - 6 - 7 548 64
          AB057411.1 - 8 - 9 649 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004099.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Rat Phosphodiesterase 1700 Inventoried Rat Phosphodiesterase Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pde7b

          Panther Classification:

          Molecular Function -

          phosphodiesterase

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          cAMP-mediated signaling
          3',5'-cyclic-AMP phosphodiesterase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P160739 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P018417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53760 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37499 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179629 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154321 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10257 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100600 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171910 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161966 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39981 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65500 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62773 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111121 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136027 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100168 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40847 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47213 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018417 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160739 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018417 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160739 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P160739 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P018417 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P102359 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101256 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35289 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26150 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47662 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75288 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18745 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17583 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30498 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73247 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90705 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93258 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60882 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10717075 - -
          Affymetrix Rat Gene 1.0 ST Array 10717077 - -
          Affymetrix Rat Gene 1.0 ST Array 10717079 - -
          Affymetrix Rat Gene 1.0 ST Array 10717082 - -
          Affymetrix Rat Gene 1.0 ST Array 10717080 - -
          Affymetrix Rat Gene 1.0 ST Array 10717081 - -
          Affymetrix Rat Gene 1.0 ST Array 10717074 - -
          Affymetrix Rat Gene 1.0 ST Array 10717072 - -
          Affymetrix Rat Gene 1.0 ST Array 10717076 - -
          Affymetrix Rat Gene 1.0 ST Array 10717078 - -
          Affymetrix Rat Gene 1.0 ST Array 10717070 - -
          Affymetrix Rat Gene 1.0 ST Array 10717071 - -
          Affymetrix Rat Gene 1.0 ST Array 10717073 - -
          Affymetrix Rat Gene 1.1 ST Array 10717069 - -
          Rat Genome 230 2.0 Array 1387582_PM_a_at - -
          Rat Genome 230 2.0 Array 1387582_a_at - -
          Affymetrix HT Rat Focus Array 1387582_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387582_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100033170.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033177.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033174.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033175.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033168.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033171.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033172.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033173.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033169.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033182.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033186.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033176.rn.1 TC0100004099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033178.rn.1 TC0100004099.rn.1 RTA TC
          Rat Expression Set 230 1387582_PM_a_at - -
          Rat Expression Set 230 1387582_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10717069 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21621687 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373020 - -
          RatRef-12 Expression BeadChip ILMN_59846 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline