Search
Search
| Gene Symbol: Trim63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080903.1 | NP_543179.1 | 2 - 3 | 431 | 56 | |
| GenBank mRNA | AY059627.1 | - | 2 - 3 | 431 | 56 | |
| BC061824.1 | - | 2 - 3 | 418 | 5599072 | 56 | |
Set Membership: |
|