Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590272_m1
          See other PPP1R14A GE Assays ›
          Gene Symbol:
          Ppp1r14a
          Gene Name:
          protein phosphatase 1, regulatory (inhibitor) subunit 14A
          Gene Aliases:
          Cpi17
          Chromosome Location:
          Chr.1: 87228581 - 87232842 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590272_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1r14a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130403.1 NP_569087.1 1 - 2 228 67
          GenBank mRNA AF352572.1 - 1 - 2 228 67
          AY050673.1 - 1 - 2 228 67
          FQ230520.1 - 1 - 2 220 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001200.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp1r14a

          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of phosphorylation
          negative regulation of catalytic activity
          protein phosphatase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P766909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59913 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P766909 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P766909 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P766909 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P766909 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10705757 - -
          Affymetrix Rat Gene 1.0 ST Array 10705754 - -
          Affymetrix Rat Gene 1.0 ST Array 10705755 - -
          Affymetrix Rat Gene 1.0 ST Array 10705756 - -
          Affymetrix Rat Gene 1.1 ST Array 10705753 - -
          Rat Genome 230 2.0 Array 1367813_PM_at - -
          Rat Genome 230 2.0 Array 1367813_at - -
          Affymetrix HT Rat Focus Array 1367813_PM_at - -
          Affymetrix HT Rat Focus Array 1367813_at - -
          Affymetrix Rat Clariom S Assay PSR0100009139.rn.1 TC0100001200.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009140.rn.1 TC0100001200.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009142.rn.1 TC0100001200.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009138.rn.1 TC0100001200.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009141.rn.1 TC0100001200.rn.1 RTA TC
          Rat Expression Set 230 1367813_at - -
          Rat Expression Set 230 1367813_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10705753 - -
          Rat Genome U34 Set rc_AI007768_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21735061 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374409 - -
          RatRef-12 Expression BeadChip ILMN_61668 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline