Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590345_m1
          See other SDF4 GE Assays ›
          Gene Symbol:
          Sdf4
          Gene Name:
          stromal cell derived factor 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 173425922 - 173444477 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590345_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sdf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130412.2 NP_569096.2 4 - 5 663 65
          XM_006239516.2 XP_006239578.1 4 - 5 711 65
          GenBank mRNA AF405545.1 - 3 - 4 573 65
          BC086996.1 - 4 - 5 663 7192272 65
          FQ218523.1 - 4 - 5 688 65
          FQ219323.1 - 4 - 5 690 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002087.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sdf4

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          UV protection
          calcium ion regulated exocytosis
          cerebellum development
          fat cell differentiation
          response to ethanol
          zymogen granule exocytosis
          calcium ion binding
          protein binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P687727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 990 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P687727 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P687727 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P687727 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P687727 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50130 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24171 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87979 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10874827 - -
          Affymetrix Rat Gene 1.0 ST Array 10874828 - -
          Affymetrix Rat Gene 1.0 ST Array 10874823 - -
          Affymetrix Rat Gene 1.0 ST Array 10874824 - -
          Affymetrix Rat Gene 1.0 ST Array 10874821 - -
          Affymetrix Rat Gene 1.0 ST Array 10874826 - -
          Affymetrix Rat Gene 1.0 ST Array 10874825 - -
          Affymetrix Rat Gene 1.0 ST Array 10874822 - -
          Affymetrix Rat Gene 1.1 ST Array 10874820 - -
          Rat Genome 230 2.0 Array 1387778_at - -
          Rat Genome 230 2.0 Array 1387778_PM_at - -
          Affymetrix HT Rat Focus Array 1387778_at - -
          Affymetrix HT Rat Focus Array 1387778_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500016060.rn.1 TC0500002087.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016066.rn.1 TC0500002087.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016069.rn.1 TC0500002087.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016070.rn.1 TC0500002087.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016061.rn.1 TC0500002087.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016065.rn.1 TC0500002087.rn.1 RTA TC
          Rat Expression Set 230 1387778_PM_at - -
          Rat Expression Set 230 1387778_at - -
          Affymetrix Rat Gene 1.0 ST Array 10874820 - -
          Rat Genome U34 Set rc_AA997861_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21740304 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354307 - -
          RatRef-12 Expression BeadChip ILMN_58801 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline