Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590358_m1
          See other SKAP2 GE Assays ›
          Gene Symbol:
          Skap2
          Gene Name:
          src kinase associated phosphoprotein 2
          Gene Aliases:
          Scap2, Scap55r
          Chromosome Location:
          Chr.4: 81816778 - 81968832 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590358_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Skap2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130413.1 NP_569097.1 9 - 10 924 73
          XM_006236477.2 XP_006236539.1 9 - 10 909 73
          XM_006236478.2 XP_006236540.1 9 - 10 909 73
          GenBank mRNA AF302132.1 - 9 - 10 924 73
          BC070949.1 - 9 - 10 871 7099436 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002890.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Skap2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s)

          negative regulation of cell proliferation
          B cell activation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108546 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112743 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1244 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59484 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119610 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47000 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100557 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22338 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142925 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33064 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159595 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75491 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13111 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13111 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13111 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13111 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6368 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93811 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23883 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52850 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48603 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 579 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32369 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63844 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86092 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24606 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103409 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10862533 - -
          Affymetrix Rat Gene 1.0 ST Array 10862540 - -
          Affymetrix Rat Gene 1.0 ST Array 10862531 - -
          Affymetrix Rat Gene 1.0 ST Array 10862534 - -
          Affymetrix Rat Gene 1.0 ST Array 10862530 - -
          Affymetrix Rat Gene 1.0 ST Array 10862539 - -
          Affymetrix Rat Gene 1.0 ST Array 10862528 - -
          Affymetrix Rat Gene 1.0 ST Array 10862538 - -
          Affymetrix Rat Gene 1.0 ST Array 10862535 - -
          Affymetrix Rat Gene 1.0 ST Array 10862537 - -
          Affymetrix Rat Gene 1.0 ST Array 10862532 - -
          Affymetrix Rat Gene 1.0 ST Array 10862529 - -
          Affymetrix Rat Gene 1.0 ST Array 10862536 - -
          Affymetrix Rat Gene 1.1 ST Array 10862527 - -
          Rat Genome 230 2.0 Array 1369979_PM_at - -
          Rat Genome 230 2.0 Array 1369979_at - -
          Affymetrix HT Rat Focus Array 1369979_PM_at - -
          Affymetrix HT Rat Focus Array 1369979_at - -
          Affymetrix Rat Clariom S Assay PSR0400020625.rn.1 TC0400002890.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020626.rn.1 TC0400002890.rn.1 RTA TC
          Rat Expression Set 230 1369979_PM_at - -
          Rat Expression Set 230 1369979_at - -
          Affymetrix Rat Gene 1.0 ST Array 10862527 - -
          Rat Genome U34 Set rc_AI010351_at - -
          Rat Genome U34 Set rc_AI232420_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22201858 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363777 - -
          RatRef-12 Expression BeadChip ILMN_60038 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline