Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590503_m1
          See other ACAA2 GE Assays ›
          Gene Symbol:
          Acaa2
          Gene Name:
          acetyl-CoA acyltransferase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 70733872 - 70762395 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590503_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acaa2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130433.1 NP_569117.1 7 - 8 1049 69
          GenBank mRNA FQ228855.1 - 7 - 8 913 69
          X05341.1 - 7 - 8 1049 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000818.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acaa2

          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          acetyl-CoA metabolic process
          fatty acid beta-oxidation
          metabolic process
          cellular response to hypoxia
          negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
          negative regulation of mitochondrial membrane permeability involved in apoptotic process
          acetyl-CoA C-acyltransferase activity
          transferase activity, transferring acyl groups other than amino-acyl groups
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P242321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804554 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157760 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147363 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P242321 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P242321 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P242321 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P242321 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P242329 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700147217 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54877 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85217 - -
          Agilent Rat cDNA Microarray (G4105A) 700147217 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802699 - -
          Affymetrix Rat Gene 1.0 ST Array 10802696 - -
          Affymetrix Rat Gene 1.0 ST Array 10802697 - -
          Affymetrix Rat Gene 1.0 ST Array 10802692 - -
          Affymetrix Rat Gene 1.0 ST Array 10802698 - -
          Affymetrix Rat Gene 1.0 ST Array 10802693 - -
          Affymetrix Rat Gene 1.0 ST Array 10802695 - -
          Affymetrix Rat Gene 1.0 ST Array 10802694 - -
          Affymetrix Rat Gene 1.1 ST Array 10802691 - -
          Rat Genome 230 2.0 Array 1386880_PM_at - -
          Rat Genome 230 2.0 Array 1380504_PM_at - -
          Rat Genome 230 2.0 Array 1386880_at - -
          Rat Genome 230 2.0 Array 1380504_at - -
          Affymetrix HT Rat Focus Array 1380504_at - -
          Affymetrix HT Rat Focus Array 1386880_PM_at - -
          Affymetrix HT Rat Focus Array 1386880_at - -
          Affymetrix HT Rat Focus Array 1380504_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800005895.rn.1 TC1800000818.rn.1 RTA TC
          Rat Expression Set 230 1386880_PM_at - -
          Rat Expression Set 230 1386880_at - -
          Rat Expression Set 230 1380504_at - -
          Rat Expression Set 230 1380504_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10802691 - -
          Rat Genome U34 Set X05341_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22007969 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357600 - -
          RatRef-12 Expression BeadChip ILMN_56900 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0