Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590612_m1
          See other LCN2 GE Assays ›
          Gene Symbol:
          Lcn2
          Gene Name:
          lipocalin 2
          Gene Aliases:
          Sip24
          Chromosome Location:
          Chr.3: 11414189 - 11417534 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590612_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lcn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130741.1 NP_570097.1 2 - 3 320 88
          GenBank mRNA BC089053.1 - 2 - 3 382 7310185 88
          X13295.1 - 2 - 3 320 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002875.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lcn2

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transport
          response to oxidative stress
          response to virus
          response to bacterium
          response to herbicide
          response to mycotoxin
          positive regulation of gene expression
          siderophore transport
          positive regulation of cell projection organization
          response to nutrient levels
          cellular response to nutrient levels
          response to drug
          innate immune response
          iron ion homeostasis
          protein homotrimerization
          cellular response to hydrogen peroxide
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          extrinsic apoptotic signaling pathway in absence of ligand
          protease binding
          transporter activity
          iron ion binding
          small molecule binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P638620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500998 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P638620 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P638620 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P638620 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P638620 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700483986 - -
          Agilent Rat cDNA Microarray (G4105A) 700483986 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10844338 - -
          Affymetrix Rat Gene 1.0 ST Array 10844334 - -
          Affymetrix Rat Gene 1.0 ST Array 10844336 - -
          Affymetrix Rat Gene 1.0 ST Array 10844335 - -
          Affymetrix Rat Gene 1.0 ST Array 10844333 - -
          Affymetrix Rat Gene 1.0 ST Array 10844337 - -
          Affymetrix Rat Gene 1.0 ST Array 10844332 - -
          Affymetrix Rat Gene 1.1 ST Array 10844331 - -
          Rat Genome 230 2.0 Array 1387011_PM_at - -
          Rat Genome 230 2.0 Array 1387011_at - -
          Affymetrix HT Rat Focus Array 1387011_at - -
          Affymetrix HT Rat Focus Array 1387011_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300022177.rn.1 TC0300002875.rn.1 RTA TC
          Rat Expression Set 230 1387011_PM_at - -
          Rat Expression Set 230 1387011_at - -
          Affymetrix Rat Gene 1.0 ST Array 10844331 - -
          Rat Genome U34 Set rc_AA946503_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21297561 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363606 - -
          RatRef-12 Expression BeadChip ILMN_68587 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline